Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 29203072-29203299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MECR
Entrez Gene ID
51102
Gene Name
mitochondrial trans-2-enoyl-CoA reductase
Gene Aliases
CGI-63, FASN2B, NRBF1
UniGene
Hs.183646
Gene Chromosome Location
Chr.1: 29192657-29230958 on Build GRCh38
Transcripts
NM_001024732.2 NM_016011.3 XM_005245885.2 XM_005245887.2 XM_011541539.1 XM_011541540.1 XM_011541541.1 XM_011541543.1 XM_011541545.1 XM_011541546.1 XM_011541547.1 XM_011541549.1 XM_011541550.1 XM_011541553.1 XM_011541554.1 XM_017001411.1 XM_017001412.1 XM_017001413.1 XM_017001414.1 XM_017001415.1 XM_017001416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748339 c.597A>G p.A199A Chr.1: 29203187-29203187 on Build GRCh38
4236514 c.604C>T p.Q202* Chr.1: 29203180-29203180 on Build GRCh38
3930740 c.114A>G p.A38A Chr.1: 29203187-29203187 on Build GRCh38
7168936 c.553_554insG p.D185fs*36 Chr.1: 29203230-29203231 on Build GRCh38
7168937 c.70_71insG p.D24fs*36 Chr.1: 29203230-29203231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140632883 Chr.1: 29203268-29203268 on Build GRCh38
rs200195789 Chr.1: 29203146-29203146 on Build GRCh38
rs752087770 Chr.1: 29203206-29203206 on Build GRCh38
rs146806836 Chr.1: 29203221-29203221 on Build GRCh38
rs1053171715 Chr.1: 29203143-29203143 on Build GRCh38
rs370277457 Chr.1: 29203160-29203160 on Build GRCh38
rs762640621 Chr.1: 29203247-29203247 on Build GRCh38
rs764985449 Chr.1: 29203199-29203199 on Build GRCh38
rs139257940 Chr.1: 29203175-29203175 on Build GRCh38
rs142970545 Chr.1: 29203202-29203202 on Build GRCh38
rs749444926 Chr.1: 29203156-29203156 on Build GRCh38
rs774524187 Chr.1: 29203265-29203265 on Build GRCh38
rs755352423 Chr.1: 29203116-29203116 on Build GRCh38
rs780633959 Chr.1: 29203241-29203241 on Build GRCh38
rs374128311 Chr.1: 29203188-29203188 on Build GRCh38
rs755442364 Chr.1: 29203215-29203215 on Build GRCh38
rs139681405 Chr.1: 29203218-29203218 on Build GRCh38
rs200619935 Chr.1: 29203201-29203201 on Build GRCh38
rs749402522 Chr.1: 29203236-29203236 on Build GRCh38
rs780747160 Chr.1: 29203122-29203122 on Build GRCh38
rs1011443165 Chr.1: 29203174-29203174 on Build GRCh38
rs867148818 Chr.1: 29203231-29203231 on Build GRCh38
rs748595179 Chr.1: 29203220-29203220 on Build GRCh38
rs778124528 Chr.1: 29203235-29203235 on Build GRCh38
rs765820789 Chr.1: 29203205-29203205 on Build GRCh38
rs770095142 Chr.1: 29203137-29203137 on Build GRCh38
rs757604329 Chr.1: 29203098-29203098 on Build GRCh38
rs998986445 Chr.1: 29203157-29203157 on Build GRCh38
rs1062924 Chr.1: 29203187-29203187 on Build GRCh38
rs149278145 Chr.1: 29203171-29203171 on Build GRCh38
rs377448817 Chr.1: 29203132-29203132 on Build GRCh38
rs375155129 Chr.1: 29203127-29203127 on Build GRCh38

View Full Product Details