Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146130902-146131163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARS
Entrez Gene ID
51520
Gene Name
leucyl-tRNA synthetase
Gene Aliases
HSPC192, ILFS1, LARS1, LEURS, LEUS, LFIS, LRS, PIG44, RNTLS, hr025Cl
UniGene
Hs.432674
Gene Chromosome Location
Chr.5: 146113026-146182787 on Build GRCh38
Transcripts
NM_001317964.1 NM_001317965.1 NM_016460.3 NM_020117.10 XM_011537656.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
134094 c.2485C>T p.Q829* Chr.5: 146131021-146131021 on Build GRCh38
1634048 c.2397-4A>G p.? Chr.5: 146131113-146131113 on Build GRCh38
1672007 c.2445G>A p.M815I Chr.5: 146131061-146131061 on Build GRCh38
1063766 c.2444T>C p.M815T Chr.5: 146131062-146131062 on Build GRCh38
6513439 c.2479G>T p.E827* Chr.5: 146131027-146131027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946827822 Chr.5: 146131065-146131065 on Build GRCh38
rs1042825288 Chr.5: 146131106-146131106 on Build GRCh38
rs754974784 Chr.5: 146130987-146130987 on Build GRCh38
rs372064359 Chr.5: 146130971-146130971 on Build GRCh38
rs774673143 Chr.5: 146131083-146131083 on Build GRCh38
rs863224046 Chr.5: 146131014-146131014 on Build GRCh38
rs142402814 Chr.5: 146130975-146130975 on Build GRCh38
rs763348707 Chr.5: 146131081-146131081 on Build GRCh38
rs375047028 Chr.5: 146130979-146130979 on Build GRCh38
rs962471796 Chr.5: 146130932-146130932 on Build GRCh38
rs536922967 Chr.5: 146130958-146130958 on Build GRCh38
rs141234474 Chr.5: 146131042-146131042 on Build GRCh38
rs763442035 Chr.5: 146130973-146130973 on Build GRCh38
rs539395367 Chr.5: 146131103-146131103 on Build GRCh38
rs756391324 Chr.5: 146131115-146131115 on Build GRCh38
rs760104325 Chr.5: 146131095-146131095 on Build GRCh38
rs767999364 Chr.5: 146131047-146131047 on Build GRCh38
rs746995255 Chr.5: 146131044-146131044 on Build GRCh38
rs369569573 Chr.5: 146131121-146131121 on Build GRCh38
rs572968230 Chr.5: 146131131-146131131 on Build GRCh38
rs752907955 Chr.5: 146130997-146130997 on Build GRCh38
rs150185886 Chr.5: 146131061-146131061 on Build GRCh38
rs778846312 Chr.5: 146131004-146131004 on Build GRCh38
rs918435700 Chr.5: 146130955-146130955 on Build GRCh38
rs553015995 Chr.5: 146131125-146131125 on Build GRCh38
rs752891357 Chr.5: 146131113-146131113 on Build GRCh38
rs941402971 Chr.5: 146131052-146131052 on Build GRCh38
rs375151931 Chr.5: 146131015-146131015 on Build GRCh38
rs766798503 Chr.5: 146131082-146131082 on Build GRCh38
rs972867102 Chr.5: 146130954-146130954 on Build GRCh38
rs200080099 Chr.5: 146130999-146130999 on Build GRCh38
rs369382242 Chr.5: 146130991-146130991 on Build GRCh38
rs745715631 Chr.5: 146131011-146131011 on Build GRCh38
rs773548613 Chr.5: 146131069-146131069 on Build GRCh38
rs776212029 Chr.5: 146131060-146131060 on Build GRCh38
rs777146277 Chr.5: 146131120-146131120 on Build GRCh38
rs769498469 Chr.5: 146131067-146131067 on Build GRCh38

View Full Product Details