Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76904271-76904496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT5B
Entrez Gene ID
146664
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B
Gene Aliases
GnT-IX, GnT-VB
UniGene
Hs.144531
Gene Chromosome Location
Chr.17: 76868487-76950389 on Build GRCh38
Transcripts
NM_001199172.1 NM_144677.2 NM_198955.1 XM_006721707.3 XM_011524350.2 XM_011524352.2 XM_011524354.2 XM_017024200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5561915 c.673C>A p.P225T Chr.17: 76904405-76904405 on Build GRCh38
5565033 c.694A>G p.R232G Chr.17: 76904393-76904393 on Build GRCh38
6081964 c.589T>C p.F197L Chr.17: 76904321-76904321 on Build GRCh38
7377094 c.689C>G p.A230G Chr.17: 76904388-76904388 on Build GRCh38
6081963 c.622T>C p.F208L Chr.17: 76904321-76904321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146591034 Chr.17: 76904401-76904401 on Build GRCh38
rs190033210 Chr.17: 76904361-76904361 on Build GRCh38
rs201242038 Chr.17: 76904311-76904311 on Build GRCh38
rs1018418061 Chr.17: 76904440-76904440 on Build GRCh38
rs753041621 Chr.17: 76904356-76904356 on Build GRCh38
rs369169563 Chr.17: 76904312-76904312 on Build GRCh38
rs543125188 Chr.17: 76904426-76904426 on Build GRCh38
rs11650151 Chr.17: 76904301-76904301 on Build GRCh38
rs761836889 Chr.17: 76904447-76904447 on Build GRCh38
rs775603524 Chr.17: 76904410-76904410 on Build GRCh38
rs760389297 Chr.17: 76904290-76904290 on Build GRCh38
rs376928919 Chr.17: 76904397-76904397 on Build GRCh38
rs140471018 Chr.17: 76904329-76904329 on Build GRCh38
rs377122452 Chr.17: 76904349-76904349 on Build GRCh38
rs999682478 Chr.17: 76904368-76904368 on Build GRCh38
rs372736073 Chr.17: 76904344-76904344 on Build GRCh38
rs778961529 Chr.17: 76904388-76904388 on Build GRCh38
rs764051103 Chr.17: 76904305-76904305 on Build GRCh38
rs746765660 Chr.17: 76904436-76904436 on Build GRCh38
rs778294137 Chr.17: 76904378-76904378 on Build GRCh38
rs903325130 Chr.17: 76904345-76904345 on Build GRCh38
rs1013237874 Chr.17: 76904430-76904430 on Build GRCh38
rs772789731 Chr.17: 76904460-76904460 on Build GRCh38
rs200272265 Chr.17: 76904446-76904446 on Build GRCh38
rs61736672 Chr.17: 76904383-76904383 on Build GRCh38
rs369519601 Chr.17: 76904382-76904382 on Build GRCh38
rs1032088398 Chr.17: 76904387-76904387 on Build GRCh38
rs775345454 Chr.17: 76904288-76904288 on Build GRCh38
rs59862466 Chr.17: 76904465-76904465 on Build GRCh38
rs772211704 Chr.17: 76904407-76904407 on Build GRCh38
rs1046291792 Chr.17: 76904364-76904364 on Build GRCh38
rs553577630 Chr.17: 76904306-76904306 on Build GRCh38
rs768310072 Chr.17: 76904438-76904438 on Build GRCh38
rs750021114 Chr.17: 76904332-76904332 on Build GRCh38

View Full Product Details