Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 225626708-225626981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYAP2
Entrez Gene ID
57624
Gene Name
neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2
Gene Aliases
KIAA1486
UniGene
Hs.224409
Gene Chromosome Location
Chr.2: 225399710-225703653 on Build GRCh38
Transcripts
NM_020864.1 XM_005246708.3 XM_011511524.1 XM_017004564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177165 c.1642C>T p.R548* Chr.2: 225626940-225626940 on Build GRCh38
4422587 c.1619-10T>G p.? Chr.2: 225626907-225626907 on Build GRCh38
5346128 c.1631A>T p.E544V Chr.2: 225626929-225626929 on Build GRCh38
5716220 c.1672T>C p.L558L Chr.2: 225626970-225626970 on Build GRCh38
4400310 c.1648C>T p.H550Y Chr.2: 225626946-225626946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989015950 Chr.2: 225626797-225626797 on Build GRCh38
rs747516864 Chr.2: 225626911-225626912 on Build GRCh38
rs1004473463 Chr.2: 225626949-225626949 on Build GRCh38
rs576312082 Chr.2: 225626847-225626847 on Build GRCh38
rs1043161851 Chr.2: 225626896-225626896 on Build GRCh38
rs754478693 Chr.2: 225626953-225626953 on Build GRCh38
rs761205121 Chr.2: 225626926-225626926 on Build GRCh38
rs772174107 Chr.2: 225626902-225626902 on Build GRCh38
rs960268184 Chr.2: 225626792-225626792 on Build GRCh38
rs61743982 Chr.2: 225626954-225626954 on Build GRCh38
rs1047829309 Chr.2: 225626913-225626913 on Build GRCh38
rs925527979 Chr.2: 225626753-225626753 on Build GRCh38
rs766009145 Chr.2: 225626940-225626940 on Build GRCh38
rs943598581 Chr.2: 225626784-225626784 on Build GRCh38
rs753357006 Chr.2: 225626941-225626941 on Build GRCh38
rs551547044 Chr.2: 225626786-225626786 on Build GRCh38
rs762138893 Chr.2: 225626919-225626919 on Build GRCh38
rs867379837 Chr.2: 225626799-225626799 on Build GRCh38
rs868825069 Chr.2: 225626923-225626923 on Build GRCh38
rs753914641 Chr.2: 225626915-225626915 on Build GRCh38
rs571658678 Chr.2: 225626789-225626789 on Build GRCh38
rs913404970 Chr.2: 225626819-225626819 on Build GRCh38
rs773856064 Chr.2: 225626924-225626924 on Build GRCh38
rs527661098 Chr.2: 225626806-225626806 on Build GRCh38
rs947511329 Chr.2: 225626865-225626865 on Build GRCh38
rs368315330 Chr.2: 225626922-225626922 on Build GRCh38

View Full Product Details