Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60927736-60927987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM132A
Entrez Gene ID
54972
Gene Name
transmembrane protein 132A
Gene Aliases
GBP, HSPA5BP1
UniGene
Hs.118552
Gene Chromosome Location
Chr.11: 60924441-60937159 on Build GRCh38
Transcripts
NM_017870.3 NM_178031.2 XM_017017951.1 XM_017017952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734248 c.416C>T p.A139V Chr.11: 60927741-60927741 on Build GRCh38
5959080 c.488delA p.H163fs*94 Chr.11: 60927813-60927813 on Build GRCh38
2036662 c.530T>G p.F177C Chr.11: 60927855-60927855 on Build GRCh38
352400 c.420G>T p.R140R Chr.11: 60927745-60927745 on Build GRCh38
929332 c.524G>A p.C175Y Chr.11: 60927849-60927849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758786158 Chr.11: 60927822-60927822 on Build GRCh38
rs775143525 Chr.11: 60927901-60927901 on Build GRCh38
rs986376500 Chr.11: 60927757-60927757 on Build GRCh38
rs576747366 Chr.11: 60927923-60927923 on Build GRCh38
rs745563168 Chr.11: 60927863-60927863 on Build GRCh38
rs781189570 Chr.11: 60927830-60927830 on Build GRCh38
rs777651889 Chr.11: 60927846-60927846 on Build GRCh38
rs770668657 Chr.11: 60927851-60927851 on Build GRCh38
rs202235322 Chr.11: 60927824-60927824 on Build GRCh38
rs559330435 Chr.11: 60927956-60927956 on Build GRCh38
rs775123362 Chr.11: 60927798-60927798 on Build GRCh38
rs1016386518 Chr.11: 60927938-60927938 on Build GRCh38
rs545657824 Chr.11: 60927924-60927924 on Build GRCh38
rs576494449 Chr.11: 60927925-60927925 on Build GRCh38
rs370154055 Chr.11: 60927779-60927779 on Build GRCh38
rs144441365 Chr.11: 60927852-60927852 on Build GRCh38
rs369462368 Chr.11: 60927837-60927837 on Build GRCh38
rs1011811778 Chr.11: 60927823-60927823 on Build GRCh38
rs1052927250 Chr.11: 60927788-60927788 on Build GRCh38
rs755894631 Chr.11: 60927967-60927967 on Build GRCh38
rs997783537 Chr.11: 60927879-60927879 on Build GRCh38
rs575955 Chr.11: 60927903-60927903 on Build GRCh38
rs928658470 Chr.11: 60927955-60927955 on Build GRCh38
rs368067544 Chr.11: 60927849-60927849 on Build GRCh38
rs970556780 Chr.11: 60927942-60927942 on Build GRCh38
rs754114057 Chr.11: 60927801-60927801 on Build GRCh38
rs753749618 Chr.11: 60927932-60927932 on Build GRCh38
rs771695533 Chr.11: 60927893-60927893 on Build GRCh38
rs980119298 Chr.11: 60927944-60927946 on Build GRCh38
rs201027775 Chr.11: 60927962-60927963 on Build GRCh38
rs554324495 Chr.11: 60927803-60927803 on Build GRCh38
rs965002361 Chr.11: 60927865-60927865 on Build GRCh38
rs374191784 Chr.11: 60927806-60927806 on Build GRCh38
rs180968764 Chr.11: 60927957-60927957 on Build GRCh38
rs773919866 Chr.11: 60927772-60927772 on Build GRCh38
rs780546391 Chr.11: 60927872-60927873 on Build GRCh38
rs575123 Chr.11: 60927829-60927829 on Build GRCh38
rs891640438 Chr.11: 60927800-60927800 on Build GRCh38
rs766999372 Chr.11: 60927793-60927793 on Build GRCh38
rs919298396 Chr.11: 60927816-60927816 on Build GRCh38
rs750847310 Chr.11: 60927821-60927821 on Build GRCh38
rs765770473 Chr.11: 60927807-60927807 on Build GRCh38
rs79330054 Chr.11: 60927962-60927962 on Build GRCh38
rs748170489 Chr.11: 60927834-60927834 on Build GRCh38
rs1019617213 Chr.11: 60927761-60927761 on Build GRCh38

View Full Product Details