Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73318587-73318834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM50
Entrez Gene ID
135892
Gene Name
tripartite motif containing 50
Gene Aliases
TRIM50A
UniGene
Hs.647053
Gene Chromosome Location
Chr.7: 73312536-73328082 on Build GRCh38
Transcripts
NM_001281450.1 NM_001281451.1 NM_178125.3 XM_011515787.1 XM_011515788.2 XM_011515789.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3086948 c.746C>G p.S249C Chr.7: 73318690-73318690 on Build GRCh38
485530 c.728A>T p.K243M Chr.7: 73318708-73318708 on Build GRCh38
4450631 c.739_749+14del25 p.? Chr.7: 73318673-73318697 on Build GRCh38
4333395 c.716A>T p.K239M Chr.7: 73318832-73318832 on Build GRCh38
3086952 c.725G>T p.R242L Chr.7: 73318823-73318823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782264211 Chr.7: 73318742-73318742 on Build GRCh38
rs782206583 Chr.7: 73318721-73318721 on Build GRCh38
rs782572305 Chr.7: 73318720-73318720 on Build GRCh38
rs781924141 Chr.7: 73318749-73318749 on Build GRCh38
rs782231174 Chr.7: 73318807-73318807 on Build GRCh38
rs782217126 Chr.7: 73318786-73318786 on Build GRCh38
rs781861674 Chr.7: 73318693-73318693 on Build GRCh38
rs543011095 Chr.7: 73318644-73318644 on Build GRCh38
rs782027912 Chr.7: 73318784-73318784 on Build GRCh38
rs782693080 Chr.7: 73318787-73318787 on Build GRCh38
rs782518450 Chr.7: 73318803-73318803 on Build GRCh38
rs782596245 Chr.7: 73318806-73318806 on Build GRCh38
rs782084216 Chr.7: 73318676-73318676 on Build GRCh38
rs782394713 Chr.7: 73318641-73318641 on Build GRCh38
rs562579048 Chr.7: 73318660-73318660 on Build GRCh38
rs782373908 Chr.7: 73318668-73318668 on Build GRCh38
rs565421644 Chr.7: 73318746-73318746 on Build GRCh38
rs774460152 Chr.7: 73318785-73318785 on Build GRCh38
rs377638120 Chr.7: 73318638-73318638 on Build GRCh38
rs375269348 Chr.7: 73318741-73318741 on Build GRCh38
rs372832421 Chr.7: 73318780-73318780 on Build GRCh38
rs371601156 Chr.7: 73318716-73318716 on Build GRCh38
rs200126161 Chr.7: 73318725-73318725 on Build GRCh38
rs536689850 Chr.7: 73318792-73318792 on Build GRCh38
rs781945671 Chr.7: 73318658-73318658 on Build GRCh38
rs369386218 Chr.7: 73318778-73318778 on Build GRCh38
rs183981056 Chr.7: 73318786-73318786 on Build GRCh38
rs771184834 Chr.7: 73318653-73318653 on Build GRCh38
rs371294385 Chr.7: 73318669-73318669 on Build GRCh38
rs199653214 Chr.7: 73318645-73318645 on Build GRCh38
rs782373414 Chr.7: 73318808-73318808 on Build GRCh38
rs782693949 Chr.7: 73318671-73318671 on Build GRCh38
rs782787112 Chr.7: 73318699-73318699 on Build GRCh38
rs782752191 Chr.7: 73318683-73318683 on Build GRCh38
rs371419477 Chr.7: 73318639-73318639 on Build GRCh38
rs782528947 Chr.7: 73318697-73318697 on Build GRCh38
rs569295101 Chr.7: 73318789-73318789 on Build GRCh38
rs532548355 Chr.7: 73318755-73318755 on Build GRCh38
rs547976639 Chr.7: 73318756-73318756 on Build GRCh38
rs782163034 Chr.7: 73318670-73318670 on Build GRCh38
rs782685197 Chr.7: 73318804-73318804 on Build GRCh38
rs782184068 Chr.7: 73318805-73318805 on Build GRCh38
rs367730941 Chr.7: 73318665-73318665 on Build GRCh38
rs782329495 Chr.7: 73318813-73318813 on Build GRCh38
rs781807166 Chr.7: 73318675-73318675 on Build GRCh38
rs559759790 Chr.7: 73318773-73318773 on Build GRCh38
rs782477015 Chr.7: 73318738-73318738 on Build GRCh38
rs782459283 Chr.7: 73318791-73318791 on Build GRCh38
rs782119309 Chr.7: 73318779-73318779 on Build GRCh38
rs192182316 Chr.7: 73318713-73318713 on Build GRCh38

View Full Product Details