Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38698055-38698292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN10A
Entrez Gene ID
6336
Gene Name
sodium voltage-gated channel alpha subunit 10
Gene Aliases
FEPS2, Nav1.8, PN3, SNS, hPN3
UniGene
Hs.250443
Gene Chromosome Location
Chr.3: 38697110-38794010 on Build GRCh38
Transcripts
NM_001293306.2 NM_001293307.2 NM_006514.3 XM_005265371.3 XM_011533993.2 XM_011533994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5617623 c.5105T>A p.F1702Y Chr.3: 38698115-38698115 on Build GRCh38
323215 c.5006C>A p.P1669H Chr.3: 38698214-38698214 on Build GRCh38
3945440 c.4956G>T p.L1652L Chr.3: 38698264-38698264 on Build GRCh38
3696069 c.4986C>T p.G1662G Chr.3: 38698234-38698234 on Build GRCh38
1309088 c.4962C>T p.L1654L Chr.3: 38698258-38698258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6599241 Chr.3: 38698083-38698083 on Build GRCh38
rs146999807 Chr.3: 38698173-38698173 on Build GRCh38
rs766952453 Chr.3: 38698123-38698123 on Build GRCh38
rs769013651 Chr.3: 38698199-38698199 on Build GRCh38
rs775273379 Chr.3: 38698237-38698237 on Build GRCh38
rs752410640 Chr.3: 38698168-38698168 on Build GRCh38
rs998456008 Chr.3: 38698241-38698241 on Build GRCh38
rs774106005 Chr.3: 38698149-38698149 on Build GRCh38
rs774384874 Chr.3: 38698218-38698218 on Build GRCh38
rs779225765 Chr.3: 38698096-38698096 on Build GRCh38
rs544529883 Chr.3: 38698240-38698240 on Build GRCh38
rs201985536 Chr.3: 38698127-38698127 on Build GRCh38
rs747204704 Chr.3: 38698138-38698138 on Build GRCh38
rs770931151 Chr.3: 38698146-38698146 on Build GRCh38
rs781206011 Chr.3: 38698200-38698200 on Build GRCh38
rs780475560 Chr.3: 38698132-38698132 on Build GRCh38
rs751007056 Chr.3: 38698262-38698262 on Build GRCh38
rs1027610429 Chr.3: 38698087-38698087 on Build GRCh38
rs759383943 Chr.3: 38698222-38698222 on Build GRCh38
rs968515082 Chr.3: 38698104-38698104 on Build GRCh38
rs758877832 Chr.3: 38698197-38698197 on Build GRCh38
rs564943632 Chr.3: 38698196-38698196 on Build GRCh38
rs147627083 Chr.3: 38698194-38698194 on Build GRCh38
rs761341190 Chr.3: 38698090-38698090 on Build GRCh38
rs769686936 Chr.3: 38698201-38698201 on Build GRCh38
rs767267531 Chr.3: 38698155-38698155 on Build GRCh38
rs112795612 Chr.3: 38698198-38698198 on Build GRCh38
rs769737839 Chr.3: 38698216-38698216 on Build GRCh38
rs150301002 Chr.3: 38698261-38698261 on Build GRCh38
rs1019979499 Chr.3: 38698270-38698270 on Build GRCh38
rs748973626 Chr.3: 38698192-38698192 on Build GRCh38
rs772552344 Chr.3: 38698191-38698192 on Build GRCh38
rs759422250 Chr.3: 38698152-38698152 on Build GRCh38
rs971950651 Chr.3: 38698160-38698160 on Build GRCh38
rs866804766 Chr.3: 38698176-38698176 on Build GRCh38
rs199541482 Chr.3: 38698119-38698119 on Build GRCh38
rs200761212 Chr.3: 38698244-38698244 on Build GRCh38
rs748280577 Chr.3: 38698213-38698213 on Build GRCh38
rs761505273 Chr.3: 38698171-38698171 on Build GRCh38
rs770506883 Chr.3: 38698213-38698213 on Build GRCh38
rs868843702 Chr.3: 38698193-38698193 on Build GRCh38
rs761742228 Chr.3: 38698143-38698143 on Build GRCh38
rs756686221 Chr.3: 38698184-38698184 on Build GRCh38
rs148256707 Chr.3: 38698264-38698264 on Build GRCh38
rs927476036 Chr.3: 38698190-38698190 on Build GRCh38
rs760392111 Chr.3: 38698239-38698239 on Build GRCh38
rs553109931 Chr.3: 38698139-38698139 on Build GRCh38
rs776838045 Chr.3: 38698141-38698141 on Build GRCh38
rs151090729 Chr.3: 38698236-38698236 on Build GRCh38
rs77804526 Chr.3: 38698131-38698131 on Build GRCh38
rs763674655 Chr.3: 38698172-38698172 on Build GRCh38
rs145756648 Chr.3: 38698142-38698142 on Build GRCh38
rs757792173 Chr.3: 38698084-38698084 on Build GRCh38
rs865835309 Chr.3: 38698227-38698227 on Build GRCh38
rs780649338 Chr.3: 38698272-38698272 on Build GRCh38

View Full Product Details