Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34739069-34739342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF181
Entrez Gene ID
339318
Gene Name
zinc finger protein 181
Gene Aliases
HHZ181
UniGene
Hs.720472
Gene Chromosome Location
Chr.19: 34733703-34746483 on Build GRCh38
Transcripts
NM_001029997.3 NM_001145665.1 XM_005258849.2 XM_005258850.2 XM_005258851.3 XM_006723183.3 XM_006723184.3 XM_017026739.1 XM_017026740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672844 c.185G>T p.W62L Chr.19: 34739191-34739191 on Build GRCh38
7106277 c.207G>C p.Q69H Chr.19: 34739213-34739213 on Build GRCh38
1612052 c.10-7A>G p.? Chr.19: 34739141-34739141 on Build GRCh38
7106276 c.75G>C p.Q25H Chr.19: 34739213-34739213 on Build GRCh38
3672843 c.53G>T p.W18L Chr.19: 34739191-34739191 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762933879 Chr.19: 34739191-34739191 on Build GRCh38
rs759863856 Chr.19: 34739279-34739279 on Build GRCh38
rs771825368 Chr.19: 34739158-34739158 on Build GRCh38
rs760537863 Chr.19: 34739170-34739170 on Build GRCh38
rs752090363 Chr.19: 34739198-34739198 on Build GRCh38
rs751006828 Chr.19: 34739318-34739318 on Build GRCh38
rs750697310 Chr.19: 34739223-34739223 on Build GRCh38
rs775560654 Chr.19: 34739304-34739304 on Build GRCh38
rs765316204 Chr.19: 34739174-34739174 on Build GRCh38
rs748680015 Chr.19: 34739137-34739137 on Build GRCh38
rs768168134 Chr.19: 34739209-34739209 on Build GRCh38
rs777698937 Chr.19: 34739286-34739286 on Build GRCh38
rs762447937 Chr.19: 34739203-34739203 on Build GRCh38
rs772955292 Chr.19: 34739169-34739169 on Build GRCh38
rs747691487 Chr.19: 34739268-34739268 on Build GRCh38
rs756508300 Chr.19: 34739226-34739226 on Build GRCh38
rs755674162 Chr.19: 34739103-34739103 on Build GRCh38
rs774261873 Chr.19: 34739142-34739142 on Build GRCh38
rs753588506 Chr.19: 34739235-34739235 on Build GRCh38
rs764325969 Chr.19: 34739193-34739193 on Build GRCh38
rs376274173 Chr.19: 34739290-34739290 on Build GRCh38
rs771945548 Chr.19: 34739274-34739274 on Build GRCh38
rs775780957 Chr.19: 34739178-34739178 on Build GRCh38
rs748137648 Chr.19: 34739151-34739151 on Build GRCh38
rs777046500 Chr.19: 34739183-34739183 on Build GRCh38
rs768071994 Chr.19: 34739315-34739315 on Build GRCh38
rs778437824 Chr.19: 34739252-34739252 on Build GRCh38
rs761998364 Chr.19: 34739313-34739313 on Build GRCh38
rs763711535 Chr.19: 34739177-34739178 on Build GRCh38
rs753116621 Chr.19: 34739309-34739309 on Build GRCh38
rs751225248 Chr.19: 34739280-34739281 on Build GRCh38
rs779569827 Chr.19: 34739228-34739228 on Build GRCh38
rs754428192 Chr.19: 34739250-34739250 on Build GRCh38
rs770822620 Chr.19: 34739303-34739303 on Build GRCh38
rs185694565 Chr.19: 34739139-34739139 on Build GRCh38
rs779772124 Chr.19: 34739114-34739114 on Build GRCh38
rs750182109 Chr.19: 34739311-34739311 on Build GRCh38

View Full Product Details