Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57398532-57398773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF548
Entrez Gene ID
147694
Gene Name
zinc finger protein 548
Gene Aliases
-
UniGene
Hs.126905
Gene Chromosome Location
Chr.19: 57389850-57403497 on Build GRCh38
Transcripts
NM_001172773.1 NM_152909.3 XM_017026316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6152630 c.346_347GG>TT p.G116L Chr.19: 57398597-57398598 on Build GRCh38
7166473 c.407A>T p.Q136L Chr.19: 57398658-57398658 on Build GRCh38
3797606 c.452C>T p.S151L Chr.19: 57398703-57398703 on Build GRCh38
2972258 c.495C>T p.F165F Chr.19: 57398746-57398746 on Build GRCh38
714210 c.304G>C p.G102R Chr.19: 57398555-57398555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200335256 Chr.19: 57398710-57398710 on Build GRCh38
rs746399068 Chr.19: 57398708-57398708 on Build GRCh38
rs763736937 Chr.19: 57398746-57398746 on Build GRCh38
rs780699822 Chr.19: 57398752-57398752 on Build GRCh38
rs765147590 Chr.19: 57398603-57398603 on Build GRCh38
rs773379443 Chr.19: 57398717-57398717 on Build GRCh38
rs769385961 Chr.19: 57398658-57398658 on Build GRCh38
rs988310120 Chr.19: 57398692-57398692 on Build GRCh38
rs749726240 Chr.19: 57398751-57398751 on Build GRCh38
rs760940894 Chr.19: 57398598-57398598 on Build GRCh38
rs751437947 Chr.19: 57398633-57398633 on Build GRCh38
rs947480851 Chr.19: 57398686-57398686 on Build GRCh38
rs758098172 Chr.19: 57398608-57398608 on Build GRCh38
rs913330837 Chr.19: 57398667-57398667 on Build GRCh38
rs769296518 Chr.19: 57398650-57398650 on Build GRCh38
rs376790357 Chr.19: 57398647-57398647 on Build GRCh38
rs752589052 Chr.19: 57398607-57398607 on Build GRCh38
rs768894353 Chr.19: 57398675-57398675 on Build GRCh38
rs1007785509 Chr.19: 57398556-57398556 on Build GRCh38
rs760816086 Chr.19: 57398720-57398720 on Build GRCh38
rs1029400960 Chr.19: 57398626-57398626 on Build GRCh38
rs767310471 Chr.19: 57398618-57398619 on Build GRCh38
rs766748095 Chr.19: 57398727-57398727 on Build GRCh38
rs199566402 Chr.19: 57398748-57398748 on Build GRCh38
rs777832943 Chr.19: 57398617-57398617 on Build GRCh38
rs1043112122 Chr.19: 57398699-57398699 on Build GRCh38
rs767440728 Chr.19: 57398582-57398582 on Build GRCh38
rs754107689 Chr.19: 57398734-57398734 on Build GRCh38
rs779654695 Chr.19: 57398659-57398659 on Build GRCh38
rs750681539 Chr.19: 57398584-57398584 on Build GRCh38
rs774938237 Chr.19: 57398706-57398706 on Build GRCh38
rs867155861 Chr.19: 57398639-57398640 on Build GRCh38
rs751517245 Chr.19: 57398747-57398747 on Build GRCh38
rs745548398 Chr.19: 57398654-57398654 on Build GRCh38
rs762863778 Chr.19: 57398739-57398739 on Build GRCh38
rs749979604 Chr.19: 57398753-57398753 on Build GRCh38
rs762015354 Chr.19: 57398573-57398573 on Build GRCh38
rs749722623 Chr.19: 57398672-57398672 on Build GRCh38

View Full Product Details