Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124322052-124322279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM65
Entrez Gene ID
157378
Gene Name
transmembrane protein 65
Gene Aliases
-
UniGene
Hs.187646
Gene Chromosome Location
Chr.8: 124306189-124372699 on Build GRCh38
Transcripts
NM_194291.2 XM_011516847.2 XM_017013104.1 XM_017013105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3896615 c.495G>T p.V165V Chr.8: 124322125-124322125 on Build GRCh38
1095856 c.490C>G p.L164V Chr.8: 124322130-124322130 on Build GRCh38
6859325 c.482_483insT p.L161fs*18 Chr.8: 124322137-124322138 on Build GRCh38
6413096 c.503T>C p.L168P Chr.8: 124322117-124322117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs80244114 Chr.8: 124322196-124322196 on Build GRCh38
rs745979993 Chr.8: 124322170-124322170 on Build GRCh38
rs371166660 Chr.8: 124322084-124322084 on Build GRCh38
rs760701143 Chr.8: 124322161-124322164 on Build GRCh38
rs918558947 Chr.8: 124322232-124322232 on Build GRCh38
rs777723097 Chr.8: 124322113-124322113 on Build GRCh38
rs778757207 Chr.8: 124322087-124322087 on Build GRCh38
rs755324171 Chr.8: 124322185-124322187 on Build GRCh38
rs1035266764 Chr.8: 124322090-124322090 on Build GRCh38
rs770299528 Chr.8: 124322171-124322171 on Build GRCh38
rs754110264 Chr.8: 124322168-124322171 on Build GRCh38
rs987150275 Chr.8: 124322235-124322235 on Build GRCh38
rs572244730 Chr.8: 124322184-124322184 on Build GRCh38
rs371759401 Chr.8: 124322193-124322193 on Build GRCh38
rs542666932 Chr.8: 124322176-124322176 on Build GRCh38
rs150119852 Chr.8: 124322201-124322201 on Build GRCh38
rs889008115 Chr.8: 124322203-124322203 on Build GRCh38
rs935395080 Chr.8: 124322162-124322162 on Build GRCh38
rs766558254 Chr.8: 124322162-124322163 on Build GRCh38
rs774766203 Chr.8: 124322197-124322197 on Build GRCh38
rs776835183 Chr.8: 124322132-124322132 on Build GRCh38
rs933988436 Chr.8: 124322233-124322233 on Build GRCh38
rs747159809 Chr.8: 124322125-124322125 on Build GRCh38
rs374599291 Chr.8: 124322089-124322089 on Build GRCh38
rs758079683 Chr.8: 124322099-124322099 on Build GRCh38
rs988031694 Chr.8: 124322168-124322168 on Build GRCh38

View Full Product Details