Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100396247-100396486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A8
Entrez Gene ID
246213
Gene Name
solute carrier family 17 member 8
Gene Aliases
DFNA25, VGLUT3
UniGene
Hs.116871
Gene Chromosome Location
Chr.12: 100357079-100422059 on Build GRCh38
Transcripts
NM_001145288.1 NM_139319.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5423949 c.637C>T p.P213S Chr.12: 100396378-100396378 on Build GRCh38
4554181 c.613G>A p.G205R Chr.12: 100396354-100396354 on Build GRCh38
4726600 c.653G>A p.R218Q Chr.12: 100396394-100396394 on Build GRCh38
1365166 c.652C>A p.R218R Chr.12: 100396393-100396393 on Build GRCh38
4037898 c.601C>A p.P201T Chr.12: 100396342-100396342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121918339 Chr.12: 100396373-100396373 on Build GRCh38
rs144949912 Chr.12: 100396308-100396308 on Build GRCh38
rs745942801 Chr.12: 100396296-100396296 on Build GRCh38
rs770924285 Chr.12: 100396425-100396425 on Build GRCh38
rs368863996 Chr.12: 100396464-100396464 on Build GRCh38
rs1037725494 Chr.12: 100396426-100396426 on Build GRCh38
rs1027059354 Chr.12: 100396462-100396462 on Build GRCh38
rs1009516241 Chr.12: 100396315-100396315 on Build GRCh38
rs1020857475 Chr.12: 100396332-100396332 on Build GRCh38
rs372770376 Chr.12: 100396435-100396435 on Build GRCh38
rs1007607527 Chr.12: 100396282-100396282 on Build GRCh38
rs780588634 Chr.12: 100396458-100396459 on Build GRCh38
rs959393397 Chr.12: 100396459-100396459 on Build GRCh38
rs775967823 Chr.12: 100396448-100396448 on Build GRCh38
rs546449115 Chr.12: 100396467-100396467 on Build GRCh38
rs753359218 Chr.12: 100396286-100396286 on Build GRCh38
rs764648553 Chr.12: 100396465-100396465 on Build GRCh38
rs774560188 Chr.12: 100396434-100396434 on Build GRCh38
rs761153997 Chr.12: 100396453-100396453 on Build GRCh38
rs772511307 Chr.12: 100396437-100396440 on Build GRCh38
rs369209552 Chr.12: 100396379-100396379 on Build GRCh38
rs944301867 Chr.12: 100396414-100396414 on Build GRCh38
rs768546214 Chr.12: 100396393-100396393 on Build GRCh38
rs776577095 Chr.12: 100396397-100396397 on Build GRCh38
rs767831162 Chr.12: 100396442-100396442 on Build GRCh38
rs147615087 Chr.12: 100396353-100396353 on Build GRCh38
rs376534425 Chr.12: 100396311-100396311 on Build GRCh38
rs756851143 Chr.12: 100396296-100396296 on Build GRCh38
rs764740432 Chr.12: 100396281-100396281 on Build GRCh38
rs11568530 Chr.12: 100396399-100396399 on Build GRCh38
rs780061011 Chr.12: 100396313-100396313 on Build GRCh38
rs868346183 Chr.12: 100396377-100396377 on Build GRCh38

View Full Product Details