Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138951699-138951954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIL1
Entrez Gene ID
64374
Gene Name
SIL1 nucleotide exchange factor
Gene Aliases
BAP, MSS, ULG5
UniGene
Hs.483521
Gene Chromosome Location
Chr.5: 138946720-139198376 on Build GRCh38
Transcripts
NM_001037633.1 NM_022464.4 XM_011543570.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3609087 c.781C>T p.Q261* Chr.5: 138951871-138951871 on Build GRCh38
4585432 c.836C>T p.T279M Chr.5: 138951816-138951816 on Build GRCh38
3850496 c.837G>A p.T279T Chr.5: 138951815-138951815 on Build GRCh38
7086832 c.796G>A p.E266K Chr.5: 138951856-138951856 on Build GRCh38
4928794 c.855A>G p.A285A Chr.5: 138951797-138951797 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752309988 Chr.5: 138951812-138951813 on Build GRCh38
rs947109793 Chr.5: 138951830-138951830 on Build GRCh38
rs921761718 Chr.5: 138951844-138951844 on Build GRCh38
rs148940232 Chr.5: 138951788-138951790 on Build GRCh38
rs769011198 Chr.5: 138951899-138951899 on Build GRCh38
rs200359743 Chr.5: 138951845-138951845 on Build GRCh38
rs758021714 Chr.5: 138951879-138951880 on Build GRCh38
rs778802859 Chr.5: 138951833-138951833 on Build GRCh38
rs372816951 Chr.5: 138951803-138951803 on Build GRCh38
rs535450021 Chr.5: 138951934-138951934 on Build GRCh38
rs776065054 Chr.5: 138951779-138951779 on Build GRCh38
rs763545682 Chr.5: 138951868-138951868 on Build GRCh38
rs201546394 Chr.5: 138951807-138951807 on Build GRCh38
rs765908702 Chr.5: 138951768-138951768 on Build GRCh38
rs532916955 Chr.5: 138951752-138951752 on Build GRCh38
rs191221829 Chr.5: 138951915-138951915 on Build GRCh38
rs376625839 Chr.5: 138951935-138951935 on Build GRCh38
rs145211892 Chr.5: 138951867-138951867 on Build GRCh38
rs753831763 Chr.5: 138951811-138951811 on Build GRCh38
rs1016026305 Chr.5: 138951856-138951856 on Build GRCh38
rs767706941 Chr.5: 138951806-138951806 on Build GRCh38
rs756645768 Chr.5: 138951782-138951783 on Build GRCh38
rs779932009 Chr.5: 138951808-138951808 on Build GRCh38
rs753325742 Chr.5: 138951745-138951745 on Build GRCh38
rs746589509 Chr.5: 138951857-138951857 on Build GRCh38
rs904069744 Chr.5: 138951784-138951784 on Build GRCh38
rs535287958 Chr.5: 138951816-138951816 on Build GRCh38
rs764540895 Chr.5: 138951791-138951791 on Build GRCh38
rs1057099472 Chr.5: 138951743-138951743 on Build GRCh38
rs914338669 Chr.5: 138951817-138951817 on Build GRCh38
rs763310911 Chr.5: 138951780-138951780 on Build GRCh38
rs760833461 Chr.5: 138951924-138951924 on Build GRCh38
rs747923562 Chr.5: 138951839-138951839 on Build GRCh38
rs377183355 Chr.5: 138951908-138951908 on Build GRCh38
rs772896004 Chr.5: 138951751-138951751 on Build GRCh38
rs986797535 Chr.5: 138951840-138951840 on Build GRCh38
rs771812516 Chr.5: 138951746-138951746 on Build GRCh38
rs750649166 Chr.5: 138951922-138951922 on Build GRCh38
rs202207368 Chr.5: 138951853-138951853 on Build GRCh38
rs770483534 Chr.5: 138951861-138951861 on Build GRCh38
rs762060202 Chr.5: 138951805-138951805 on Build GRCh38
rs995660522 Chr.5: 138951757-138951757 on Build GRCh38
rs373574853 Chr.5: 138951930-138951930 on Build GRCh38
rs1035913513 Chr.5: 138951815-138951815 on Build GRCh38

View Full Product Details