Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20967832-20968091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4102812 c.82G>A p.A28T Chr.22: 20967952-20967952 on Build GRCh38
6597182 c.23C>A p.P8H Chr.22: 20967967-20967967 on Build GRCh38
4102814 c.29C>T p.P10L Chr.22: 20967973-20967973 on Build GRCh38
6320923 c.46G>T p.A16S Chr.22: 20967916-20967916 on Build GRCh38
4102811 c.8G>A p.G3D Chr.22: 20967952-20967952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774770417 Chr.22: 20967999-20967999 on Build GRCh38
rs377510436 Chr.22: 20968037-20968037 on Build GRCh38
rs894765855 Chr.22: 20967931-20967931 on Build GRCh38
rs1002318221 Chr.22: 20967911-20967911 on Build GRCh38
rs371215112 Chr.22: 20967942-20967942 on Build GRCh38
rs764274218 Chr.22: 20967857-20967857 on Build GRCh38
rs566902132 Chr.22: 20967959-20967959 on Build GRCh38
rs749674814 Chr.22: 20967974-20967974 on Build GRCh38
rs865989947 Chr.22: 20967892-20967892 on Build GRCh38
rs528647446 Chr.22: 20967880-20967880 on Build GRCh38
rs773465429 Chr.22: 20968019-20968019 on Build GRCh38
rs746544875 Chr.22: 20967909-20967909 on Build GRCh38
rs772827076 Chr.22: 20967855-20967855 on Build GRCh38
rs796877852 Chr.22: 20968045-20968045 on Build GRCh38
rs777487675 Chr.22: 20967946-20967946 on Build GRCh38
rs781282442 Chr.22: 20967873-20967873 on Build GRCh38
rs546346131 Chr.22: 20968044-20968044 on Build GRCh38
rs767825991 Chr.22: 20968002-20968002 on Build GRCh38
rs765233522 Chr.22: 20967941-20967941 on Build GRCh38
rs1006222906 Chr.22: 20968031-20968031 on Build GRCh38
rs768833316 Chr.22: 20967986-20967986 on Build GRCh38
rs1036923709 Chr.22: 20968061-20968061 on Build GRCh38
rs374136982 Chr.22: 20967956-20967956 on Build GRCh38
rs976751419 Chr.22: 20968033-20968033 on Build GRCh38
rs760070671 Chr.22: 20967859-20967859 on Build GRCh38
rs746707375 Chr.22: 20967949-20967949 on Build GRCh38
rs757449518 Chr.22: 20967872-20967873 on Build GRCh38
rs777066125 Chr.22: 20967861-20967861 on Build GRCh38
rs747934158 Chr.22: 20967893-20967893 on Build GRCh38
rs755014255 Chr.22: 20967939-20967939 on Build GRCh38
rs1026414306 Chr.22: 20968013-20968013 on Build GRCh38
rs975229686 Chr.22: 20968045-20968045 on Build GRCh38
rs866271982 Chr.22: 20968042-20968042 on Build GRCh38
rs538411353 Chr.22: 20968051-20968051 on Build GRCh38
rs753050681 Chr.22: 20967872-20967872 on Build GRCh38
rs372042715 Chr.22: 20967920-20967920 on Build GRCh38
rs964177133 Chr.22: 20967850-20967850 on Build GRCh38
rs771609239 Chr.22: 20967851-20967851 on Build GRCh38
rs909782533 Chr.22: 20967874-20967874 on Build GRCh38
rs201078355 Chr.22: 20967923-20967923 on Build GRCh38
rs367705545 Chr.22: 20968023-20968023 on Build GRCh38
rs778909558 Chr.22: 20967882-20967882 on Build GRCh38
rs966701047 Chr.22: 20968006-20968006 on Build GRCh38
rs545199522 Chr.22: 20967922-20967922 on Build GRCh38
rs770280649 Chr.22: 20967910-20967910 on Build GRCh38
rs774375673 Chr.22: 20967934-20967934 on Build GRCh38
rs761995501 Chr.22: 20968000-20968000 on Build GRCh38
rs376941423 Chr.22: 20968046-20968046 on Build GRCh38
rs941210587 Chr.22: 20968060-20968060 on Build GRCh38
rs767768802 Chr.22: 20967938-20967938 on Build GRCh38
rs933756158 Chr.22: 20967869-20967869 on Build GRCh38
rs759899458 Chr.22: 20967863-20967863 on Build GRCh38
rs761954077 Chr.22: 20967937-20967937 on Build GRCh38
rs1017259830 Chr.22: 20968039-20968039 on Build GRCh38
rs952109251 Chr.22: 20968015-20968015 on Build GRCh38
rs780050452 Chr.22: 20967879-20967879 on Build GRCh38
rs558725184 Chr.22: 20967862-20967862 on Build GRCh38
rs1055079152 Chr.22: 20967928-20967928 on Build GRCh38
rs529727715 Chr.22: 20968034-20968034 on Build GRCh38
rs751844786 Chr.22: 20967856-20967857 on Build GRCh38
rs11334069 Chr.22: 20968038-20968038 on Build GRCh38
rs922679770 Chr.22: 20968043-20968043 on Build GRCh38
rs771795525 Chr.22: 20967902-20967902 on Build GRCh38
rs866861978 Chr.22: 20967908-20967908 on Build GRCh38
rs765657412 Chr.22: 20967864-20967864 on Build GRCh38
rs764194057 Chr.22: 20967875-20967875 on Build GRCh38
rs780734241 Chr.22: 20967951-20967951 on Build GRCh38
rs896796446 Chr.22: 20967891-20967891 on Build GRCh38
rs566584544 Chr.22: 20968050-20968050 on Build GRCh38
rs763288838 Chr.22: 20967873-20967873 on Build GRCh38
rs148333657 Chr.22: 20967950-20967950 on Build GRCh38
rs922452720 Chr.22: 20968047-20968047 on Build GRCh38
rs760093700 Chr.22: 20967913-20967913 on Build GRCh38
rs751693537 Chr.22: 20967878-20967878 on Build GRCh38
rs777248634 Chr.22: 20967903-20967903 on Build GRCh38
rs146335337 Chr.22: 20967955-20967955 on Build GRCh38
rs178254 Chr.22: 20968063-20968063 on Build GRCh38

View Full Product Details