Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114350598-114350823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB20
Entrez Gene ID
26137
Gene Name
zinc finger and BTB domain containing 20
Gene Aliases
DPZF, HOF, ODA-8S, PRIMS, ZNF288
UniGene
Hs.202577
Gene Chromosome Location
Chr.3: 114314500-115147280 on Build GRCh38
Transcripts
NM_001164342.2 NM_001164343.2 NM_001164344.2 NM_001164345.2 NM_001164346.2 NM_001164347.2 NM_015642.5 NR_121662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6825097 c.1357G>A p.D453N Chr.3: 114350721-114350721 on Build GRCh38
3119496 c.1310A>G p.N437S Chr.3: 114350768-114350768 on Build GRCh38
3119497 c.1091A>G p.N364S Chr.3: 114350768-114350768 on Build GRCh38
6825105 c.1345A>C p.S449R Chr.3: 114350733-114350733 on Build GRCh38
5663775 c.1149C>T p.V383V Chr.3: 114350710-114350710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756725802 Chr.3: 114350746-114350746 on Build GRCh38
rs145364827 Chr.3: 114350712-114350712 on Build GRCh38
rs770278746 Chr.3: 114350689-114350689 on Build GRCh38
rs1016200753 Chr.3: 114350671-114350671 on Build GRCh38
rs149179361 Chr.3: 114350755-114350755 on Build GRCh38
rs750873192 Chr.3: 114350737-114350737 on Build GRCh38
rs540652273 Chr.3: 114350677-114350677 on Build GRCh38
rs138924453 Chr.3: 114350760-114350760 on Build GRCh38
rs897150333 Chr.3: 114350683-114350683 on Build GRCh38
rs754310386 Chr.3: 114350756-114350756 on Build GRCh38
rs140765510 Chr.3: 114350674-114350674 on Build GRCh38
rs957893205 Chr.3: 114350793-114350793 on Build GRCh38
rs776479104 Chr.3: 114350695-114350695 on Build GRCh38
rs998700576 Chr.3: 114350687-114350687 on Build GRCh38
rs746920213 Chr.3: 114350803-114350803 on Build GRCh38
rs763151962 Chr.3: 114350696-114350696 on Build GRCh38
rs777387926 Chr.3: 114350799-114350799 on Build GRCh38
rs754743360 Chr.3: 114350632-114350632 on Build GRCh38
rs532683480 Chr.3: 114350792-114350792 on Build GRCh38
rs778711177 Chr.3: 114350779-114350779 on Build GRCh38
rs868811407 Chr.3: 114350638-114350638 on Build GRCh38
rs766845109 Chr.3: 114350747-114350747 on Build GRCh38
rs778996878 Chr.3: 114350645-114350645 on Build GRCh38
rs748917483 Chr.3: 114350629-114350629 on Build GRCh38
rs780910146 Chr.3: 114350806-114350806 on Build GRCh38
rs954212230 Chr.3: 114350750-114350750 on Build GRCh38
rs752208031 Chr.3: 114350780-114350780 on Build GRCh38
rs1006124149 Chr.3: 114350627-114350627 on Build GRCh38
rs780614361 Chr.3: 114350625-114350625 on Build GRCh38
rs768021143 Chr.3: 114350717-114350717 on Build GRCh38
rs747623097 Chr.3: 114350646-114350646 on Build GRCh38
rs377216471 Chr.3: 114350640-114350640 on Build GRCh38
rs770686482 Chr.3: 114350680-114350680 on Build GRCh38
rs184110076 Chr.3: 114350713-114350713 on Build GRCh38
rs970841722 Chr.3: 114350761-114350761 on Build GRCh38

View Full Product Details