Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104759387-104759612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIVA1
Entrez Gene ID
10572
Gene Name
SIVA1 apoptosis inducing factor
Gene Aliases
CD27BP, SIVA, Siva-1, Siva-2
UniGene
Hs.112058
Gene Chromosome Location
Chr.14: 104752420-104759659 on Build GRCh38
Transcripts
NM_006427.3 NM_021709.2 XM_011536360.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
292089 c.490A>T p.K164* Chr.14: 104759447-104759447 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766361338 Chr.14: 104759533-104759533 on Build GRCh38
rs370091956 Chr.14: 104759486-104759486 on Build GRCh38
rs771423066 Chr.14: 104759521-104759521 on Build GRCh38
rs779844712 Chr.14: 104759423-104759423 on Build GRCh38
rs913919354 Chr.14: 104759547-104759547 on Build GRCh38
rs762694387 Chr.14: 104759463-104759463 on Build GRCh38
rs376249415 Chr.14: 104759513-104759513 on Build GRCh38
rs14905 Chr.14: 104759551-104759551 on Build GRCh38
rs910160519 Chr.14: 104759583-104759583 on Build GRCh38
rs750275814 Chr.14: 104759477-104759477 on Build GRCh38
rs763900745 Chr.14: 104759468-104759468 on Build GRCh38
rs747149970 Chr.14: 104759444-104759444 on Build GRCh38
rs766093404 Chr.14: 104759484-104759484 on Build GRCh38
rs755823032 Chr.14: 104759421-104759421 on Build GRCh38
rs773980647 Chr.14: 104759471-104759471 on Build GRCh38
rs767399938 Chr.14: 104759409-104759409 on Build GRCh38
rs149750921 Chr.14: 104759476-104759476 on Build GRCh38
rs757236168 Chr.14: 104759497-104759497 on Build GRCh38
rs778018804 Chr.14: 104759491-104759491 on Build GRCh38
rs781346755 Chr.14: 104759499-104759499 on Build GRCh38
rs939433826 Chr.14: 104759522-104759522 on Build GRCh38
rs553915968 Chr.14: 104759417-104759417 on Build GRCh38
rs770492092 Chr.14: 104759506-104759506 on Build GRCh38
rs375749455 Chr.14: 104759452-104759452 on Build GRCh38
rs749722984 Chr.14: 104759512-104759512 on Build GRCh38
rs139437432 Chr.14: 104759455-104759455 on Build GRCh38
rs756088094 Chr.14: 104759481-104759481 on Build GRCh38
rs945272947 Chr.14: 104759548-104759548 on Build GRCh38
rs1039194179 Chr.14: 104759580-104759580 on Build GRCh38
rs903671280 Chr.14: 104759552-104759552 on Build GRCh38
rs901556140 Chr.14: 104759581-104759581 on Build GRCh38
rs376097428 Chr.14: 104759536-104759536 on Build GRCh38
rs771437927 Chr.14: 104759449-104759449 on Build GRCh38
rs774049267 Chr.14: 104759438-104759439 on Build GRCh38
rs185050950 Chr.14: 104759525-104759525 on Build GRCh38
rs559184664 Chr.14: 104759514-104759514 on Build GRCh38
rs373057611 Chr.14: 104759501-104759501 on Build GRCh38
rs758605452 Chr.14: 104759488-104759488 on Build GRCh38
rs371919593 Chr.14: 104759424-104759424 on Build GRCh38
rs150511651 Chr.14: 104759439-104759439 on Build GRCh38
rs539489780 Chr.14: 104759510-104759510 on Build GRCh38
rs746410201 Chr.14: 104759454-104759454 on Build GRCh38
rs369632432 Chr.14: 104759517-104759517 on Build GRCh38
rs1032294130 Chr.14: 104759466-104759466 on Build GRCh38
rs202092612 Chr.14: 104759443-104759443 on Build GRCh38
rs981256096 Chr.14: 104759500-104759500 on Build GRCh38
rs755781334 Chr.14: 104759584-104759584 on Build GRCh38
rs760524308 Chr.14: 104759529-104759529 on Build GRCh38
rs943253471 Chr.14: 104759556-104759556 on Build GRCh38

View Full Product Details