Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 479843-480072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100288152
Entrez Gene ID
100288152
Gene Name
uncharacterized LOC100288152
Gene Aliases
-
UniGene
Hs.535801
Gene Chromosome Location
Chr.5: 473236-480892 on Build GRCh38
Transcripts
NR_125375.1 NM_001284351.2 NM_004174.3

Gene Symbol
SLC9A3
Entrez Gene ID
6550
Gene Name
solute carrier family 9 member A3
Gene Aliases
DIAR8, NHE-3, NHE3
UniGene
Hs.658120
Gene Chromosome Location
Chr.5: 471986-524434 on Build GRCh38
Transcripts
NR_125375.1 NM_001284351.2 NM_004174.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5811301 c.1618C>A p.L540M Chr.5: 479865-479865 on Build GRCh38
7166789 c.1620G>A p.L540L Chr.5: 479863-479863 on Build GRCh38
6783716 c.1639G>A p.V547M Chr.5: 479844-479844 on Build GRCh38
5451028 c.1583G>A p.R528Q Chr.5: 479900-479900 on Build GRCh38
461976 c.1545C>T p.L515L Chr.5: 479938-479938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879138547 Chr.5: 479979-479979 on Build GRCh38
rs1006770414 Chr.5: 480021-480021 on Build GRCh38
rs997452344 Chr.5: 480005-480005 on Build GRCh38
rs767813750 Chr.5: 479957-479957 on Build GRCh38
rs948301970 Chr.5: 479908-479908 on Build GRCh38
rs759835293 Chr.5: 479865-479865 on Build GRCh38
rs199904762 Chr.5: 480018-480018 on Build GRCh38
rs748096261 Chr.5: 480010-480010 on Build GRCh38
rs750763652 Chr.5: 479998-479998 on Build GRCh38
rs371820443 Chr.5: 479874-479874 on Build GRCh38
rs781082547 Chr.5: 480007-480007 on Build GRCh38
rs1017305696 Chr.5: 480022-480022 on Build GRCh38
rs535304229 Chr.5: 480039-480039 on Build GRCh38
rs778170621 Chr.5: 479884-479884 on Build GRCh38
rs1011516819 Chr.5: 479919-479919 on Build GRCh38
rs754319359 Chr.5: 479887-479887 on Build GRCh38
rs571029514 Chr.5: 480027-480027 on Build GRCh38
rs144147429 Chr.5: 479909-479909 on Build GRCh38
rs999269945 Chr.5: 479907-479907 on Build GRCh38
rs761745958 Chr.5: 479931-479931 on Build GRCh38
rs745585295 Chr.5: 479896-479896 on Build GRCh38
rs147340203 Chr.5: 479914-479914 on Build GRCh38
rs752924063 Chr.5: 479963-479963 on Build GRCh38
rs754303649 Chr.5: 479986-479986 on Build GRCh38
rs960422263 Chr.5: 480034-480034 on Build GRCh38
rs369450763 Chr.5: 479953-479953 on Build GRCh38
rs780752424 Chr.5: 480009-480009 on Build GRCh38
rs767767021 Chr.5: 479872-479872 on Build GRCh38
rs139467260 Chr.5: 479918-479918 on Build GRCh38
rs559219255 Chr.5: 480026-480026 on Build GRCh38
rs187798401 Chr.5: 480025-480025 on Build GRCh38
rs769546146 Chr.5: 479952-479952 on Build GRCh38
rs757572851 Chr.5: 479894-479894 on Build GRCh38
rs756084605 Chr.5: 480028-480028 on Build GRCh38
rs769836881 Chr.5: 480015-480015 on Build GRCh38
rs758724020 Chr.5: 480002-480002 on Build GRCh38
rs201095264 Chr.5: 480006-480006 on Build GRCh38
rs759890286 Chr.5: 479956-479956 on Build GRCh38
rs376396781 Chr.5: 479900-479900 on Build GRCh38
rs764173072 Chr.5: 479975-479975 on Build GRCh38
rs183706873 Chr.5: 480004-480004 on Build GRCh38
rs757775831 Chr.5: 479993-479993 on Build GRCh38
rs756680787 Chr.5: 479875-479875 on Build GRCh38
rs367993737 Chr.5: 479869-479869 on Build GRCh38
rs779180547 Chr.5: 479895-479895 on Build GRCh38
rs369382043 Chr.5: 479968-479968 on Build GRCh38
rs779580593 Chr.5: 479906-479906 on Build GRCh38
rs779307295 Chr.5: 479996-479996 on Build GRCh38

View Full Product Details