Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73978158-73978405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCP2
Entrez Gene ID
7351
Gene Name
uncoupling protein 2
Gene Aliases
BMIQ4, SLC25A8, UCPH
UniGene
Hs.80658
Gene Chromosome Location
Chr.11: 73974671-73982844 on Build GRCh38
Transcripts
NM_003355.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5607191 c.97C>T p.P33S Chr.11: 73978282-73978282 on Build GRCh38
4828798 c.59C>A p.A20D Chr.11: 73978320-73978320 on Build GRCh38
1243717 c.126G>T p.Q42H Chr.11: 73978253-73978253 on Build GRCh38
2017210 c.118C>T p.R40W Chr.11: 73978261-73978261 on Build GRCh38
195855 c.79G>A p.A27T Chr.11: 73978300-73978300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374415462 Chr.11: 73978383-73978383 on Build GRCh38
rs142733319 Chr.11: 73978371-73978371 on Build GRCh38
rs755177729 Chr.11: 73978303-73978303 on Build GRCh38
rs926061114 Chr.11: 73978366-73978366 on Build GRCh38
rs147381195 Chr.11: 73978378-73978378 on Build GRCh38
rs374054689 Chr.11: 73978348-73978348 on Build GRCh38
rs369586591 Chr.11: 73978220-73978220 on Build GRCh38
rs764113779 Chr.11: 73978357-73978357 on Build GRCh38
rs747409980 Chr.11: 73978326-73978326 on Build GRCh38
rs377720978 Chr.11: 73978261-73978261 on Build GRCh38
rs765451373 Chr.11: 73978384-73978384 on Build GRCh38
rs761674109 Chr.11: 73978363-73978363 on Build GRCh38
rs370781460 Chr.11: 73978356-73978356 on Build GRCh38
rs776204470 Chr.11: 73978259-73978259 on Build GRCh38
rs774480323 Chr.11: 73978320-73978320 on Build GRCh38
rs771130610 Chr.11: 73978332-73978332 on Build GRCh38
rs770652835 Chr.11: 73978257-73978257 on Build GRCh38
rs758911603 Chr.11: 73978309-73978309 on Build GRCh38
rs779568764 Chr.11: 73978213-73978213 on Build GRCh38
rs45574134 Chr.11: 73978260-73978260 on Build GRCh38
rs762852709 Chr.11: 73978263-73978263 on Build GRCh38
rs775461544 Chr.11: 73978262-73978262 on Build GRCh38
rs774097794 Chr.11: 73978361-73978361 on Build GRCh38
rs571993764 Chr.11: 73978201-73978201 on Build GRCh38
rs746684870 Chr.11: 73978252-73978252 on Build GRCh38
rs772380898 Chr.11: 73978242-73978242 on Build GRCh38
rs148253207 Chr.11: 73978264-73978264 on Build GRCh38
rs745503729 Chr.11: 73978345-73978345 on Build GRCh38
rs569544010 Chr.11: 73978308-73978308 on Build GRCh38
rs754173175 Chr.11: 73978379-73978379 on Build GRCh38
rs755264006 Chr.11: 73978299-73978299 on Build GRCh38
rs774948044 Chr.11: 73978354-73978354 on Build GRCh38
rs370639209 Chr.11: 73978294-73978294 on Build GRCh38
rs756953904 Chr.11: 73978275-73978275 on Build GRCh38
rs45532033 Chr.11: 73978207-73978207 on Build GRCh38
rs773607480 Chr.11: 73978245-73978245 on Build GRCh38
rs771149390 Chr.11: 73978216-73978216 on Build GRCh38
rs779124511 Chr.11: 73978306-73978306 on Build GRCh38
rs752859030 Chr.11: 73978385-73978385 on Build GRCh38
rs749401120 Chr.11: 73978184-73978184 on Build GRCh38
rs780098901 Chr.11: 73978181-73978181 on Build GRCh38
rs201315561 Chr.11: 73978300-73978300 on Build GRCh38
rs755051243 Chr.11: 73978203-73978203 on Build GRCh38
rs756328495 Chr.11: 73978178-73978178 on Build GRCh38
rs776329175 Chr.11: 73978334-73978334 on Build GRCh38

View Full Product Details