Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24465560-24465831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7337090 c.352G>A p.D118N Chr.8: 24465738-24465738 on Build GRCh38
142116 c.316C>T p.H106Y Chr.8: 24465702-24465702 on Build GRCh38
4971191 c.374C>A p.T125K Chr.8: 24465760-24465760 on Build GRCh38
3834458 c.369C>A p.I123I Chr.8: 24465755-24465755 on Build GRCh38
5912741 c.348A>T p.E116D Chr.8: 24465734-24465734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028615062 Chr.8: 24465793-24465793 on Build GRCh38
rs764671483 Chr.8: 24465676-24465676 on Build GRCh38
rs940085461 Chr.8: 24465732-24465732 on Build GRCh38
rs746374813 Chr.8: 24465730-24465730 on Build GRCh38
rs142219351 Chr.8: 24465729-24465729 on Build GRCh38
rs754099694 Chr.8: 24465777-24465777 on Build GRCh38
rs924415063 Chr.8: 24465619-24465619 on Build GRCh38
rs938610331 Chr.8: 24465625-24465625 on Build GRCh38
rs767236507 Chr.8: 24465760-24465760 on Build GRCh38
rs767980101 Chr.8: 24465657-24465657 on Build GRCh38
rs1050833220 Chr.8: 24465635-24465635 on Build GRCh38
rs751500124 Chr.8: 24465755-24465755 on Build GRCh38
rs147639395 Chr.8: 24465706-24465706 on Build GRCh38
rs749903492 Chr.8: 24465685-24465685 on Build GRCh38
rs968442405 Chr.8: 24465594-24465594 on Build GRCh38
rs115629541 Chr.8: 24465612-24465612 on Build GRCh38
rs758018970 Chr.8: 24465733-24465733 on Build GRCh38
rs373237334 Chr.8: 24465719-24465719 on Build GRCh38
rs201780677 Chr.8: 24465662-24465662 on Build GRCh38
rs766349335 Chr.8: 24465691-24465691 on Build GRCh38
rs140158202 Chr.8: 24465761-24465761 on Build GRCh38
rs376045894 Chr.8: 24465759-24465759 on Build GRCh38
rs777746470 Chr.8: 24465723-24465723 on Build GRCh38
rs369075452 Chr.8: 24465774-24465774 on Build GRCh38
rs567709882 Chr.8: 24465690-24465690 on Build GRCh38
rs992724529 Chr.8: 24465661-24465661 on Build GRCh38
rs771113387 Chr.8: 24465726-24465726 on Build GRCh38
rs775868912 Chr.8: 24465734-24465734 on Build GRCh38
rs34017992 Chr.8: 24465731-24465731 on Build GRCh38
rs762838914 Chr.8: 24465724-24465724 on Build GRCh38
rs762504011 Chr.8: 24465741-24465741 on Build GRCh38
rs761465689 Chr.8: 24465672-24465672 on Build GRCh38
rs752875517 Chr.8: 24465714-24465714 on Build GRCh38
rs373413048 Chr.8: 24465780-24465780 on Build GRCh38
rs142907504 Chr.8: 24465738-24465738 on Build GRCh38
rs200135966 Chr.8: 24465711-24465711 on Build GRCh38
rs185121881 Chr.8: 24465736-24465736 on Build GRCh38
rs267601867 Chr.8: 24465702-24465702 on Build GRCh38
rs150302349 Chr.8: 24465770-24465770 on Build GRCh38
rs780372349 Chr.8: 24465677-24465678 on Build GRCh38
rs761396783 Chr.8: 24465735-24465735 on Build GRCh38
rs751481647 Chr.8: 24465692-24465692 on Build GRCh38
rs765864524 Chr.8: 24465744-24465744 on Build GRCh38

View Full Product Details