Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76736936-76737171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
UniGene
Hs.732019
Gene Chromosome Location
Chr.17: 76736565-76803805 on Build GRCh38
Transcripts
NM_001242534.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Gene Symbol
SRSF2
Entrez Gene ID
6427
Gene Name
serine and arginine rich splicing factor 2
Gene Aliases
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
UniGene
Hs.584801
Gene Chromosome Location
Chr.17: 76734115-76737431 on Build GRCh38
Transcripts
NM_001242534.1 NM_001195427.1 NM_003016.4 NR_036608.1 XM_017024942.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390702 c.156G>C p.K52N Chr.17: 76737005-76737005 on Build GRCh38
6961994 c.35G>A p.G12D Chr.17: 76737126-76737126 on Build GRCh38
375315 c.39G>C p.M13I Chr.17: 76737122-76737122 on Build GRCh38
6494513 c.75C>T p.T25T Chr.17: 76737086-76737086 on Build GRCh38
6965783 c.157G>A p.E53K Chr.17: 76737004-76737004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756403208 Chr.17: 76737098-76737098 on Build GRCh38
rs770956483 Chr.17: 76737005-76737005 on Build GRCh38
rs237057 Chr.17: 76737017-76737017 on Build GRCh38
rs778600867 Chr.17: 76736969-76736969 on Build GRCh38
rs17849462 Chr.17: 76737116-76737116 on Build GRCh38
rs763988734 Chr.17: 76737047-76737047 on Build GRCh38
rs61742084 Chr.17: 76737086-76737086 on Build GRCh38
rs142294903 Chr.17: 76736993-76736993 on Build GRCh38
rs372496093 Chr.17: 76737065-76737065 on Build GRCh38
rs140957080 Chr.17: 76737080-76737080 on Build GRCh38
rs774991189 Chr.17: 76737131-76737131 on Build GRCh38
rs11550963 Chr.17: 76737022-76737022 on Build GRCh38
rs546284289 Chr.17: 76737119-76737119 on Build GRCh38
rs775513335 Chr.17: 76737146-76737146 on Build GRCh38
rs999131144 Chr.17: 76737044-76737044 on Build GRCh38
rs774394478 Chr.17: 76737008-76737008 on Build GRCh38
rs779792829 Chr.17: 76736987-76736987 on Build GRCh38
rs773650940 Chr.17: 76736999-76736999 on Build GRCh38
rs376379121 Chr.17: 76736990-76736990 on Build GRCh38
rs200354490 Chr.17: 76736978-76736978 on Build GRCh38
rs1031504488 Chr.17: 76737062-76737062 on Build GRCh38
rs745477669 Chr.17: 76736974-76736974 on Build GRCh38
rs1009427086 Chr.17: 76737029-76737029 on Build GRCh38
rs757286065 Chr.17: 76737054-76737054 on Build GRCh38
rs760991597 Chr.17: 76737149-76737149 on Build GRCh38
rs778139770 Chr.17: 76737104-76737104 on Build GRCh38
rs756763573 Chr.17: 76736960-76736960 on Build GRCh38
rs148089239 Chr.17: 76737074-76737074 on Build GRCh38
rs61742139 Chr.17: 76737038-76737038 on Build GRCh38
rs775840051 Chr.17: 76737028-76737028 on Build GRCh38
rs772206470 Chr.17: 76737143-76737143 on Build GRCh38
rs745923097 Chr.17: 76737137-76737137 on Build GRCh38
rs575538276 Chr.17: 76737050-76737050 on Build GRCh38
rs369871019 Chr.17: 76736991-76736991 on Build GRCh38
rs759702439 Chr.17: 76737014-76737014 on Build GRCh38
rs757990285 Chr.17: 76737077-76737077 on Build GRCh38

View Full Product Details