Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 146939621-146939860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC29
Entrez Gene ID
83894
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
NYD-SP14, TBPP2A
UniGene
Hs.378893
Gene Chromosome Location
Chr.4: 146706638-146945886 on Build GRCh38
Transcripts
NM_001300761.2 NM_001317806.1 NM_031956.3 NR_133922.1 XM_005263270.1 XM_006714335.1 XM_006714336.1 XM_006714339.2 XM_011532310.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5400595 c.56G>A p.R19K Chr.4: 146939840-146939840 on Build GRCh38
5400594 c.134G>A p.R45K Chr.4: 146939840-146939840 on Build GRCh38
7395585 c.38A>C p.K13T Chr.4: 146939858-146939858 on Build GRCh38
4786360 c.153C>A p.S51S Chr.4: 146939821-146939821 on Build GRCh38
5737110 c.154T>C p.S52P Chr.4: 146939820-146939820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755210021 Chr.4: 146939786-146939786 on Build GRCh38
rs576373246 Chr.4: 146939700-146939700 on Build GRCh38
rs1015201244 Chr.4: 146939827-146939827 on Build GRCh38
rs901812134 Chr.4: 146939774-146939774 on Build GRCh38
rs61741876 Chr.4: 146939828-146939828 on Build GRCh38
rs779670641 Chr.4: 146939664-146939664 on Build GRCh38
rs751635860 Chr.4: 146939784-146939784 on Build GRCh38
rs867749590 Chr.4: 146939807-146939807 on Build GRCh38
rs763156501 Chr.4: 146939772-146939772 on Build GRCh38
rs778536285 Chr.4: 146939800-146939800 on Build GRCh38
rs556410900 Chr.4: 146939658-146939658 on Build GRCh38
rs753633608 Chr.4: 146939667-146939667 on Build GRCh38
rs781469396 Chr.4: 146939790-146939790 on Build GRCh38
rs756985091 Chr.4: 146939794-146939794 on Build GRCh38
rs370817128 Chr.4: 146939783-146939783 on Build GRCh38
rs753575411 Chr.4: 146939793-146939793 on Build GRCh38
rs771978293 Chr.4: 146939806-146939806 on Build GRCh38
rs1000156912 Chr.4: 146939740-146939740 on Build GRCh38
rs776289710 Chr.4: 146939829-146939829 on Build GRCh38
rs779491976 Chr.4: 146939808-146939808 on Build GRCh38
rs377435583 Chr.4: 146939754-146939754 on Build GRCh38
rs974038723 Chr.4: 146939686-146939686 on Build GRCh38
rs902749551 Chr.4: 146939674-146939674 on Build GRCh38
rs59050226 Chr.4: 146939799-146939799 on Build GRCh38
rs950104045 Chr.4: 146939823-146939823 on Build GRCh38
rs746526382 Chr.4: 146939811-146939811 on Build GRCh38

View Full Product Details