Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114099073-114099300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT6
Entrez Gene ID
148281
Gene Name
synaptotagmin 6
Gene Aliases
sytVI
UniGene
Hs.370963
Gene Chromosome Location
Chr.1: 114089292-114153919 on Build GRCh38
Transcripts
NM_001270805.1 NM_205848.3 XM_017000371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6835246 c.1193-1G>C p.? Chr.1: 114099266-114099266 on Build GRCh38
6864207 c.1289A>T p.E430V Chr.1: 114099169-114099169 on Build GRCh38
201654 c.1059G>A p.P353P Chr.1: 114099144-114099144 on Build GRCh38
6896944 c.1346C>T p.S449L Chr.1: 114099112-114099112 on Build GRCh38
6795547 c.1098G>T p.M366I Chr.1: 114099105-114099105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748547825 Chr.1: 114099168-114099168 on Build GRCh38
rs751002681 Chr.1: 114099128-114099128 on Build GRCh38
rs201961031 Chr.1: 114099199-114099199 on Build GRCh38
rs780597166 Chr.1: 114099144-114099144 on Build GRCh38
rs889443495 Chr.1: 114099244-114099244 on Build GRCh38
rs749981191 Chr.1: 114099250-114099250 on Build GRCh38
rs78377200 Chr.1: 114099213-114099213 on Build GRCh38
rs377580497 Chr.1: 114099243-114099243 on Build GRCh38
rs772366530 Chr.1: 114099173-114099173 on Build GRCh38
rs764976999 Chr.1: 114099092-114099092 on Build GRCh38
rs766978733 Chr.1: 114099249-114099249 on Build GRCh38
rs769744385 Chr.1: 114099225-114099225 on Build GRCh38
rs762741956 Chr.1: 114099229-114099229 on Build GRCh38
rs867514138 Chr.1: 114099149-114099149 on Build GRCh38
rs374072146 Chr.1: 114099122-114099122 on Build GRCh38
rs773143623 Chr.1: 114099106-114099106 on Build GRCh38
rs149899868 Chr.1: 114099095-114099095 on Build GRCh38
rs751271416 Chr.1: 114099239-114099239 on Build GRCh38
rs779383865 Chr.1: 114099146-114099146 on Build GRCh38
rs200088199 Chr.1: 114099230-114099230 on Build GRCh38
rs966022120 Chr.1: 114099237-114099237 on Build GRCh38
rs775205575 Chr.1: 114099226-114099226 on Build GRCh38
rs953213849 Chr.1: 114099112-114099112 on Build GRCh38
rs370506806 Chr.1: 114099275-114099275 on Build GRCh38
rs767345262 Chr.1: 114099216-114099216 on Build GRCh38
rs759349402 Chr.1: 114099099-114099099 on Build GRCh38
rs1051262035 Chr.1: 114099155-114099155 on Build GRCh38
rs770961129 Chr.1: 114099212-114099212 on Build GRCh38
rs752518998 Chr.1: 114099094-114099094 on Build GRCh38
rs753047173 Chr.1: 114099276-114099276 on Build GRCh38
rs147172888 Chr.1: 114099258-114099258 on Build GRCh38
rs779581001 Chr.1: 114099274-114099274 on Build GRCh38
rs757994972 Chr.1: 114099119-114099119 on Build GRCh38
rs551632114 Chr.1: 114099203-114099203 on Build GRCh38
rs200649940 Chr.1: 114099145-114099145 on Build GRCh38
rs773576074 Chr.1: 114099188-114099188 on Build GRCh38
rs756784538 Chr.1: 114099137-114099137 on Build GRCh38

View Full Product Details