Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 122918755-122919016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB26
Entrez Gene ID
57684
Gene Name
zinc finger and BTB domain containing 26
Gene Aliases
ZNF481, bioref
UniGene
Hs.5638
Gene Chromosome Location
Chr.9: 122918029-122931551 on Build GRCh38
Transcripts
NM_001304363.1 NM_001304364.1 NM_020924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3903881 c.995G>A p.G332E Chr.9: 122918940-122918940 on Build GRCh38
3324019 c.1098A>G p.K366K Chr.9: 122918837-122918837 on Build GRCh38
4742118 c.1089delT p.F363fs*7 Chr.9: 122918846-122918846 on Build GRCh38
6020766 c.1161G>C p.E387D Chr.9: 122918774-122918774 on Build GRCh38
3324017 c.1129C>A p.H377N Chr.9: 122918806-122918806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867207603 Chr.9: 122918956-122918956 on Build GRCh38
rs780959531 Chr.9: 122918990-122918990 on Build GRCh38
rs750406557 Chr.9: 122918876-122918876 on Build GRCh38
rs754803858 Chr.9: 122918981-122918981 on Build GRCh38
rs202128661 Chr.9: 122918795-122918795 on Build GRCh38
rs371291127 Chr.9: 122918972-122918972 on Build GRCh38
rs766253217 Chr.9: 122918911-122918911 on Build GRCh38
rs761932184 Chr.9: 122918783-122918783 on Build GRCh38
rs747891706 Chr.9: 122918993-122918993 on Build GRCh38
rs377088624 Chr.9: 122918906-122918906 on Build GRCh38

View Full Product Details