Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78932251-78932502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSH
Entrez Gene ID
1512
Gene Name
cathepsin H
Gene Aliases
ACC-4, ACC-5, ACC4, ACC5, CPSB
UniGene
Hs.148641
Gene Chromosome Location
Chr.15: 78921750-78945098 on Build GRCh38
Transcripts
NM_001319137.1 NM_004390.4 XM_005254181.2 XM_011521276.1 XM_017021951.1 XM_017021952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
965443 c.432C>A p.F144L Chr.15: 78932432-78932432 on Build GRCh38
4057180 c.414C>T p.C138C Chr.15: 78932450-78932450 on Build GRCh38
6637812 c.422G>A p.C141Y Chr.15: 78932442-78932442 on Build GRCh38
3504377 c.488C>T p.S163F Chr.15: 78932376-78932376 on Build GRCh38
6995690 c.418A>C p.S140R Chr.15: 78932446-78932446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752857196 Chr.15: 78932472-78932472 on Build GRCh38
rs141613289 Chr.15: 78932407-78932407 on Build GRCh38
rs149231812 Chr.15: 78932390-78932390 on Build GRCh38
rs765326132 Chr.15: 78932387-78932387 on Build GRCh38
rs886037648 Chr.15: 78932376-78932379 on Build GRCh38
rs944705766 Chr.15: 78932368-78932368 on Build GRCh38
rs747505120 Chr.15: 78932396-78932396 on Build GRCh38
rs771415886 Chr.15: 78932327-78932327 on Build GRCh38
rs1033300744 Chr.15: 78932441-78932441 on Build GRCh38
rs776428294 Chr.15: 78932450-78932450 on Build GRCh38
rs767532192 Chr.15: 78932357-78932357 on Build GRCh38
rs773846126 Chr.15: 78932410-78932410 on Build GRCh38
rs370693086 Chr.15: 78932332-78932332 on Build GRCh38
rs147068368 Chr.15: 78932416-78932416 on Build GRCh38
rs902281536 Chr.15: 78932434-78932434 on Build GRCh38
rs749842358 Chr.15: 78932424-78932424 on Build GRCh38
rs750358310 Chr.15: 78932361-78932361 on Build GRCh38
rs573645982 Chr.15: 78932342-78932342 on Build GRCh38
rs2289700 Chr.15: 78932341-78932341 on Build GRCh38
rs758377577 Chr.15: 78932389-78932389 on Build GRCh38
rs760524676 Chr.15: 78932364-78932364 on Build GRCh38
rs745687624 Chr.15: 78932419-78932419 on Build GRCh38
rs747764581 Chr.15: 78932411-78932411 on Build GRCh38
rs200021038 Chr.15: 78932360-78932360 on Build GRCh38
rs759226585 Chr.15: 78932452-78932452 on Build GRCh38
rs930511039 Chr.15: 78932333-78932333 on Build GRCh38
rs374675140 Chr.15: 78932461-78932461 on Build GRCh38
rs13345 Chr.15: 78932405-78932405 on Build GRCh38
rs752868085 Chr.15: 78932388-78932388 on Build GRCh38
rs78155742 Chr.15: 78932385-78932385 on Build GRCh38
rs753566686 Chr.15: 78932383-78932383 on Build GRCh38
rs773934583 Chr.15: 78932343-78932343 on Build GRCh38
rs758561179 Chr.15: 78932476-78932476 on Build GRCh38
rs375198024 Chr.15: 78932376-78932376 on Build GRCh38
rs775340404 Chr.15: 78932337-78932337 on Build GRCh38
rs78513281 Chr.15: 78932444-78932444 on Build GRCh38
rs143214964 Chr.15: 78932401-78932401 on Build GRCh38
rs781714615 Chr.15: 78932402-78932402 on Build GRCh38
rs369287227 Chr.15: 78932352-78932352 on Build GRCh38
rs747417195 Chr.15: 78932321-78932321 on Build GRCh38
rs766261285 Chr.15: 78932449-78932449 on Build GRCh38
rs147587459 Chr.15: 78932288-78932288 on Build GRCh38
rs760614809 Chr.15: 78932421-78932421 on Build GRCh38

View Full Product Details