Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76588791-76589036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001285460.1 NM_006633.3 XM_005248410.2 XM_017008960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1070193 c.470T>C p.F157S Chr.5: 76588917-76588917 on Build GRCh38
5964257 c.471C>A p.F157L Chr.5: 76588918-76588918 on Build GRCh38
5501001 c.526+10T>G p.? Chr.5: 76588983-76588983 on Build GRCh38
1438621 c.506T>C p.L169S Chr.5: 76588953-76588953 on Build GRCh38
7117291 c.516A>G p.V172V Chr.5: 76588963-76588963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013335645 Chr.5: 76588827-76588827 on Build GRCh38
rs764630385 Chr.5: 76588891-76588892 on Build GRCh38
rs749170197 Chr.5: 76588864-76588864 on Build GRCh38
rs1015904819 Chr.5: 76588914-76588914 on Build GRCh38
rs752998022 Chr.5: 76588940-76588940 on Build GRCh38
rs775114361 Chr.5: 76588861-76588861 on Build GRCh38
rs754137958 Chr.5: 76588920-76588920 on Build GRCh38
rs745812498 Chr.5: 76588857-76588857 on Build GRCh38
rs773999780 Chr.5: 76588873-76588873 on Build GRCh38
rs777974408 Chr.5: 76588973-76588973 on Build GRCh38
rs181492978 Chr.5: 76589003-76589003 on Build GRCh38
rs772829085 Chr.5: 76588899-76588899 on Build GRCh38
rs761277064 Chr.5: 76588882-76588882 on Build GRCh38
rs767040531 Chr.5: 76588889-76588889 on Build GRCh38
rs202016097 Chr.5: 76588934-76588934 on Build GRCh38
rs758500626 Chr.5: 76588964-76588964 on Build GRCh38
rs375045201 Chr.5: 76588925-76588925 on Build GRCh38
rs747150538 Chr.5: 76588996-76588996 on Build GRCh38
rs761278890 Chr.5: 76588892-76588892 on Build GRCh38
rs371558272 Chr.5: 76588859-76588859 on Build GRCh38
rs768243004 Chr.5: 76588866-76588866 on Build GRCh38
rs757645782 Chr.5: 76588996-76588996 on Build GRCh38
rs755382504 Chr.5: 76588922-76588922 on Build GRCh38
rs1131232 Chr.5: 76588909-76588909 on Build GRCh38
rs766978308 Chr.5: 76588911-76588911 on Build GRCh38

View Full Product Details