Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89013968-89014223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP3
Entrez Gene ID
2635
Gene Name
guanylate binding protein 3
Gene Aliases
-
UniGene
Hs.720167
Gene Chromosome Location
Chr.1: 89006677-89022880 on Build GRCh38
Transcripts
NM_001319179.1 NM_001319180.1 NM_001319181.1 NM_018284.2 NR_135007.1 XM_005270743.2 XM_006710574.2 XM_011541233.1 XM_011541234.1 XM_017001013.1 XM_017001014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686549 c.522A>G p.P174P Chr.1: 89014186-89014186 on Build GRCh38
3806054 c.577C>A p.P193T Chr.1: 89014131-89014131 on Build GRCh38
912944 c.549C>A p.F183L Chr.1: 89014159-89014159 on Build GRCh38
6219801 c.586C>A p.P196T Chr.1: 89014122-89014122 on Build GRCh38
912945 c.542G>T p.R181I Chr.1: 89014166-89014166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747309715 Chr.1: 89014057-89014057 on Build GRCh38
rs371737476 Chr.1: 89014051-89014051 on Build GRCh38
rs1021063709 Chr.1: 89014173-89014173 on Build GRCh38
rs921244667 Chr.1: 89014006-89014006 on Build GRCh38
rs749859692 Chr.1: 89014168-89014168 on Build GRCh38
rs977193989 Chr.1: 89014137-89014137 on Build GRCh38
rs984030549 Chr.1: 89014018-89014018 on Build GRCh38
rs767150621 Chr.1: 89014082-89014082 on Build GRCh38
rs746254195 Chr.1: 89014124-89014124 on Build GRCh38
rs184102383 Chr.1: 89014016-89014016 on Build GRCh38
rs747446049 Chr.1: 89014084-89014084 on Build GRCh38
rs371269489 Chr.1: 89014068-89014068 on Build GRCh38
rs909808264 Chr.1: 89014050-89014050 on Build GRCh38
rs756051773 Chr.1: 89014047-89014047 on Build GRCh38
rs3795543 Chr.1: 89014075-89014075 on Build GRCh38
rs770434829 Chr.1: 89014070-89014070 on Build GRCh38
rs143690907 Chr.1: 89014115-89014115 on Build GRCh38
rs772244380 Chr.1: 89014054-89014054 on Build GRCh38
rs777116554 Chr.1: 89014062-89014062 on Build GRCh38
rs770383566 Chr.1: 89014130-89014130 on Build GRCh38
rs200573819 Chr.1: 89014088-89014088 on Build GRCh38
rs991823295 Chr.1: 89014087-89014087 on Build GRCh38
rs377188594 Chr.1: 89014056-89014056 on Build GRCh38
rs781639673 Chr.1: 89014122-89014122 on Build GRCh38
rs866838818 Chr.1: 89014108-89014108 on Build GRCh38
rs551463714 Chr.1: 89014003-89014003 on Build GRCh38
rs771183787 Chr.1: 89014060-89014060 on Build GRCh38
rs750356169 Chr.1: 89014037-89014037 on Build GRCh38
rs768037958 Chr.1: 89014171-89014171 on Build GRCh38
rs993643548 Chr.1: 89014014-89014014 on Build GRCh38
rs761594458 Chr.1: 89014078-89014078 on Build GRCh38
rs763700614 Chr.1: 89014074-89014074 on Build GRCh38
rs755998362 Chr.1: 89014101-89014101 on Build GRCh38
rs567658856 Chr.1: 89014065-89014065 on Build GRCh38
rs560581644 Chr.1: 89014004-89014004 on Build GRCh38
rs776107022 Chr.1: 89014157-89014157 on Build GRCh38
rs375143324 Chr.1: 89014107-89014107 on Build GRCh38
rs772954243 Chr.1: 89014099-89014099 on Build GRCh38
rs144891343 Chr.1: 89014183-89014183 on Build GRCh38
rs775090421 Chr.1: 89014071-89014071 on Build GRCh38
rs766337281 Chr.1: 89014104-89014104 on Build GRCh38
rs12091541 Chr.1: 89014072-89014072 on Build GRCh38
rs778908132 Chr.1: 89014048-89014048 on Build GRCh38
rs757501381 Chr.1: 89014117-89014117 on Build GRCh38

View Full Product Details