Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159907190-159907457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAS1
Entrez Gene ID
4142
Gene Name
MAS1 proto-oncogene, G protein-coupled receptor
Gene Aliases
MAS, MGRA
UniGene
Hs.99900
Gene Chromosome Location
Chr.6: 159906942-159908076 on Build GRCh38
Transcripts
NM_002377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1075573 c.365T>C p.L122P Chr.6: 159907320-159907320 on Build GRCh38
3622467 c.354G>A p.T118T Chr.6: 159907309-159907309 on Build GRCh38
94678 c.358C>T p.L120F Chr.6: 159907313-159907313 on Build GRCh38
88599 c.353C>T p.T118M Chr.6: 159907308-159907308 on Build GRCh38
5003750 c.494C>A p.T165N Chr.6: 159907449-159907449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753527518 Chr.6: 159907386-159907386 on Build GRCh38
rs754192055 Chr.6: 159907300-159907300 on Build GRCh38
rs889218376 Chr.6: 159907274-159907274 on Build GRCh38
rs372604345 Chr.6: 159907247-159907247 on Build GRCh38
rs767448750 Chr.6: 159907255-159907255 on Build GRCh38
rs532916357 Chr.6: 159907329-159907329 on Build GRCh38
rs142220585 Chr.6: 159907326-159907326 on Build GRCh38
rs768531368 Chr.6: 159907327-159907327 on Build GRCh38
rs770809893 Chr.6: 159907218-159907218 on Build GRCh38
rs375732408 Chr.6: 159907389-159907389 on Build GRCh38
rs773393107 Chr.6: 159907435-159907435 on Build GRCh38
rs761022711 Chr.6: 159907287-159907287 on Build GRCh38
rs756810150 Chr.6: 159907312-159907312 on Build GRCh38
rs780051704 Chr.6: 159907324-159907324 on Build GRCh38
rs541092772 Chr.6: 159907284-159907284 on Build GRCh38
rs765532855 Chr.6: 159907385-159907385 on Build GRCh38
rs759077969 Chr.6: 159907229-159907234 on Build GRCh38
rs371805366 Chr.6: 159907306-159907306 on Build GRCh38
rs763695360 Chr.6: 159907336-159907336 on Build GRCh38
rs762016372 Chr.6: 159907248-159907248 on Build GRCh38
rs759921715 Chr.6: 159907376-159907376 on Build GRCh38
rs756790658 Chr.6: 159907387-159907387 on Build GRCh38
rs145329855 Chr.6: 159907418-159907418 on Build GRCh38
rs145130069 Chr.6: 159907348-159907348 on Build GRCh38
rs745741176 Chr.6: 159907222-159907222 on Build GRCh38
rs865921787 Chr.6: 159907364-159907364 on Build GRCh38
rs749189737 Chr.6: 159907408-159907408 on Build GRCh38
rs754835270 Chr.6: 159907409-159907409 on Build GRCh38
rs773163149 Chr.6: 159907342-159907342 on Build GRCh38
rs761885116 Chr.6: 159907330-159907330 on Build GRCh38
rs772173297 Chr.6: 159907430-159907430 on Build GRCh38
rs768986845 Chr.6: 159907244-159907244 on Build GRCh38
rs75396648 Chr.6: 159907278-159907278 on Build GRCh38
rs145079703 Chr.6: 159907308-159907308 on Build GRCh38
rs780506777 Chr.6: 159907314-159907314 on Build GRCh38
rs761306044 Chr.6: 159907345-159907345 on Build GRCh38
rs769642224 Chr.6: 159907223-159907223 on Build GRCh38
rs140242572 Chr.6: 159907357-159907357 on Build GRCh38
rs139649334 Chr.6: 159907259-159907259 on Build GRCh38
rs751168993 Chr.6: 159907309-159907309 on Build GRCh38
rs763332083 Chr.6: 159907242-159907242 on Build GRCh38
rs748472255 Chr.6: 159907427-159907427 on Build GRCh38
rs1007803275 Chr.6: 159907282-159907282 on Build GRCh38
rs190132211 Chr.6: 159907402-159907402 on Build GRCh38
rs749886003 Chr.6: 159907395-159907395 on Build GRCh38
rs149100513 Chr.6: 159907323-159907323 on Build GRCh38
rs745819582 Chr.6: 159907318-159907318 on Build GRCh38
rs747248947 Chr.6: 159907401-159907401 on Build GRCh38
rs149942969 Chr.6: 159907299-159907299 on Build GRCh38
rs755508039 Chr.6: 159907236-159907236 on Build GRCh38
rs781217007 Chr.6: 159907221-159907221 on Build GRCh38

View Full Product Details