Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144212891-144213152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH1
Entrez Gene ID
727957
Gene Name
maestro heat like repeat family member 1
Gene Aliases
HEATR7A
UniGene
Hs.443139
Gene Chromosome Location
Chr.8: 144148016-144261940 on Build GRCh38
Transcripts
NM_001099280.2 NM_001099281.2 NM_001288814.1 NM_032450.2 XM_011517270.2 XM_011517271.2 XM_011517272.2 XM_011517273.2 XM_011517274.2 XM_011517275.2 XM_011517276.2 XM_011517277.1 XM_011517278.1 XM_011517279.2 XM_011517281.1 XM_011517282.1 XM_011517283.2 XM_011517285.1 XM_011517286.2 XM_017013802.1 XM_017013803.1 XM_017013804.1 XM_017013805.1 XM_017013806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2870986 c.1216C>T p.R406C Chr.8: 144213068-144213068 on Build GRCh38
1455593 c.1248C>T p.A416A Chr.8: 144213100-144213100 on Build GRCh38
6715103 c.1233T>C p.P411P Chr.8: 144213085-144213085 on Build GRCh38
2870989 c.1257G>A p.A419A Chr.8: 144213109-144213109 on Build GRCh38
5997894 c.1145G>A p.S382N Chr.8: 144212997-144212997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758677715 Chr.8: 144212969-144212969 on Build GRCh38
rs373612134 Chr.8: 144213101-144213101 on Build GRCh38
rs758910610 Chr.8: 144213117-144213117 on Build GRCh38
rs74345248 Chr.8: 144213106-144213106 on Build GRCh38
rs781421052 Chr.8: 144212987-144212988 on Build GRCh38
rs768407136 Chr.8: 144213078-144213078 on Build GRCh38
rs749225406 Chr.8: 144212998-144212998 on Build GRCh38
rs749248379 Chr.8: 144213099-144213099 on Build GRCh38
rs917466781 Chr.8: 144212949-144212949 on Build GRCh38
rs375703350 Chr.8: 144212962-144212962 on Build GRCh38
rs747060861 Chr.8: 144212977-144212977 on Build GRCh38
rs755350985 Chr.8: 144212950-144212950 on Build GRCh38
rs928242571 Chr.8: 144213019-144213019 on Build GRCh38
rs760086075 Chr.8: 144213064-144213064 on Build GRCh38
rs746286131 Chr.8: 144213020-144213021 on Build GRCh38
rs950206046 Chr.8: 144212961-144212961 on Build GRCh38
rs369400734 Chr.8: 144213100-144213100 on Build GRCh38
rs376778532 Chr.8: 144213041-144213041 on Build GRCh38
rs371234349 Chr.8: 144213129-144213129 on Build GRCh38
rs368703338 Chr.8: 144213068-144213068 on Build GRCh38
rs916765423 Chr.8: 144213062-144213062 on Build GRCh38
rs776193934 Chr.8: 144213109-144213109 on Build GRCh38
rs753122567 Chr.8: 144213069-144213069 on Build GRCh38
rs532310066 Chr.8: 144212934-144212934 on Build GRCh38
rs761540699 Chr.8: 144213028-144213028 on Build GRCh38
rs1006637298 Chr.8: 144212930-144212930 on Build GRCh38
rs746268878 Chr.8: 144213008-144213008 on Build GRCh38
rs757329215 Chr.8: 144213096-144213096 on Build GRCh38
rs201651665 Chr.8: 144213102-144213102 on Build GRCh38
rs374781788 Chr.8: 144212923-144212923 on Build GRCh38
rs774126655 Chr.8: 144213027-144213027 on Build GRCh38
rs373393781 Chr.8: 144213079-144213079 on Build GRCh38
rs12114251 Chr.8: 144213097-144213097 on Build GRCh38
rs565953387 Chr.8: 144213018-144213018 on Build GRCh38
rs770417479 Chr.8: 144213108-144213108 on Build GRCh38
rs753036954 Chr.8: 144212964-144212964 on Build GRCh38
rs369801015 Chr.8: 144212971-144212971 on Build GRCh38
rs779878794 Chr.8: 144212984-144212984 on Build GRCh38
rs973913829 Chr.8: 144212958-144212958 on Build GRCh38
rs371210609 Chr.8: 144213103-144213103 on Build GRCh38
rs376468888 Chr.8: 144213107-144213107 on Build GRCh38
rs370101798 Chr.8: 144213130-144213130 on Build GRCh38
rs772813232 Chr.8: 144213042-144213042 on Build GRCh38
rs964796562 Chr.8: 144212945-144212945 on Build GRCh38
rs751756547 Chr.8: 144213080-144213080 on Build GRCh38
rs111350180 Chr.8: 144212980-144212980 on Build GRCh38
rs765895358 Chr.8: 144213050-144213050 on Build GRCh38
rs754346261 Chr.8: 144213055-144213055 on Build GRCh38

View Full Product Details