Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99352194-99352427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC28
Entrez Gene ID
123355
Gene Name
leucine rich repeat containing 28
Gene Aliases
-
UniGene
Hs.578684
Gene Chromosome Location
Chr.15: 99251362-99390732 on Build GRCh38
Transcripts
NM_001284400.2 NM_001321675.1 NM_001321676.1 NM_001321677.1 NM_001321678.1 NM_001321679.1 NM_001321680.1 NM_144598.4 NR_135753.1 NR_135754.1 NR_135755.1 NR_135756.1 NR_135757.1 NR_135758.1 NR_135759.1 XM_006720389.3 XM_011521218.2 XM_011521220.1 XM_011521221.2 XM_017021914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850124 c.599G>T p.G200V Chr.15: 99352375-99352375 on Build GRCh38
1375635 c.638A>G p.N213S Chr.15: 99352414-99352414 on Build GRCh38
1708584 c.607C>T p.R203* Chr.15: 99352383-99352383 on Build GRCh38
4995628 c.625T>C p.Y209H Chr.15: 99352401-99352401 on Build GRCh38
966892 c.628G>A p.V210M Chr.15: 99352404-99352404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567748631 Chr.15: 99352317-99352317 on Build GRCh38
rs1004948276 Chr.15: 99352298-99352298 on Build GRCh38
rs770083132 Chr.15: 99352320-99352320 on Build GRCh38
rs368442873 Chr.15: 99352299-99352299 on Build GRCh38
rs772006097 Chr.15: 99352387-99352387 on Build GRCh38
rs762329853 Chr.15: 99352348-99352348 on Build GRCh38
rs907903257 Chr.15: 99352300-99352300 on Build GRCh38
rs781137940 Chr.15: 99352391-99352391 on Build GRCh38
rs754810054 Chr.15: 99352378-99352378 on Build GRCh38
rs566718516 Chr.15: 99352305-99352305 on Build GRCh38
rs951234974 Chr.15: 99352290-99352290 on Build GRCh38
rs949423200 Chr.15: 99352278-99352278 on Build GRCh38
rs973851424 Chr.15: 99352332-99352332 on Build GRCh38
rs1015450053 Chr.15: 99352312-99352312 on Build GRCh38
rs1039619817 Chr.15: 99352233-99352233 on Build GRCh38
rs368069445 Chr.15: 99352377-99352377 on Build GRCh38
rs375122144 Chr.15: 99352336-99352336 on Build GRCh38
rs958236007 Chr.15: 99352218-99352218 on Build GRCh38
rs551776888 Chr.15: 99352277-99352277 on Build GRCh38
rs963402475 Chr.15: 99352315-99352315 on Build GRCh38
rs766962685 Chr.15: 99352353-99352353 on Build GRCh38
rs1029420403 Chr.15: 99352286-99352286 on Build GRCh38
rs777043254 Chr.15: 99352364-99352364 on Build GRCh38
rs991359689 Chr.15: 99352236-99352236 on Build GRCh38
rs139516247 Chr.15: 99352384-99352384 on Build GRCh38
rs940663737 Chr.15: 99352303-99352303 on Build GRCh38
rs775841239 Chr.15: 99352321-99352321 on Build GRCh38
rs1046497919 Chr.15: 99352292-99352292 on Build GRCh38
rs778658681 Chr.15: 99352383-99352383 on Build GRCh38
rs763475605 Chr.15: 99352334-99352334 on Build GRCh38
rs969592800 Chr.15: 99352369-99352369 on Build GRCh38
rs775894135 Chr.15: 99352400-99352400 on Build GRCh38
rs899801051 Chr.15: 99352267-99352267 on Build GRCh38
rs746017164 Chr.15: 99352396-99352396 on Build GRCh38
rs749878665 Chr.15: 99352361-99352361 on Build GRCh38
rs113136321 Chr.15: 99352284-99352284 on Build GRCh38
rs192444767 Chr.15: 99352335-99352335 on Build GRCh38

View Full Product Details