Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67254830-67255071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521915.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Gene Symbol
IQCH
Entrez Gene ID
64799
Gene Name
IQ motif containing H
Gene Aliases
NYDSP5
UniGene
Hs.657894
Gene Chromosome Location
Chr.15: 67254772-67501804 on Build GRCh38
Transcripts
NM_001271885.1 NM_001031715.2 NM_001284347.1 NM_001284348.1 NM_001284349.1 NM_001322470.1 NM_001322471.1 NM_001322472.1 NM_001322473.1 NM_001322474.1 NM_001322475.1 NM_022784.2 XM_011521907.2 XM_011521908.2 XM_011521911.2 XM_011521912.2 XM_011521913.2 XM_011521915.2 XM_011521918.2 XM_011521922.2 XM_011521923.2 XM_017022491.1 XM_017022492.1 XM_017022493.1 XM_017022494.1 XM_017022495.1 XM_017022496.1 XM_017022497.1 XM_017022498.1 XM_017022499.1 XM_017022500.1 XM_017022501.1 XM_017022502.1 XM_017022503.1 XM_017022504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363084 c.15T>C p.T5T Chr.15: 67254911-67254911 on Build GRCh38
1210855 c.45A>C p.L15F Chr.15: 67254941-67254941 on Build GRCh38
5971510 c.51+1G>A p.? Chr.15: 67254948-67254948 on Build GRCh38
1210856 c.45A>C p.L15F Chr.15: 67254941-67254941 on Build GRCh38
2257451 c.7C>T p.Q3* Chr.15: 67254903-67254903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999039916 Chr.15: 67255022-67255022 on Build GRCh38
rs937732358 Chr.15: 67254881-67254881 on Build GRCh38
rs370216523 Chr.15: 67254872-67254872 on Build GRCh38
rs952337276 Chr.15: 67254999-67254999 on Build GRCh38
rs769739034 Chr.15: 67254926-67254926 on Build GRCh38
rs770868761 Chr.15: 67254990-67254990 on Build GRCh38
rs774136973 Chr.15: 67254947-67254947 on Build GRCh38
rs376484021 Chr.15: 67254878-67254878 on Build GRCh38
rs3743348 Chr.15: 67255011-67255011 on Build GRCh38
rs371799427 Chr.15: 67254997-67254997 on Build GRCh38
rs762543048 Chr.15: 67254850-67254850 on Build GRCh38
rs903375728 Chr.15: 67255019-67255019 on Build GRCh38
rs939941137 Chr.15: 67254875-67254875 on Build GRCh38
rs3743347 Chr.15: 67254963-67254963 on Build GRCh38
rs777735907 Chr.15: 67254929-67254929 on Build GRCh38
rs113435495 Chr.15: 67254857-67254857 on Build GRCh38
rs775030011 Chr.15: 67254950-67254950 on Build GRCh38
rs758655425 Chr.15: 67254871-67254871 on Build GRCh38
rs750789718 Chr.15: 67254904-67254904 on Build GRCh38
rs751994186 Chr.15: 67254968-67254968 on Build GRCh38
rs555020732 Chr.15: 67255051-67255051 on Build GRCh38
rs901456764 Chr.15: 67255028-67255028 on Build GRCh38
rs757709854 Chr.15: 67254867-67254867 on Build GRCh38
rs781306979 Chr.15: 67254972-67254972 on Build GRCh38
rs867372021 Chr.15: 67254916-67254916 on Build GRCh38
rs903428420 Chr.15: 67254901-67254901 on Build GRCh38
rs766048528 Chr.15: 67254851-67254851 on Build GRCh38
rs752444584 Chr.15: 67254868-67254868 on Build GRCh38
rs533073588 Chr.15: 67254974-67254974 on Build GRCh38
rs754435742 Chr.15: 67254863-67254863 on Build GRCh38
rs762775024 Chr.15: 67254885-67254885 on Build GRCh38
rs559840295 Chr.15: 67254900-67254900 on Build GRCh38
rs752166368 Chr.15: 67254897-67254897 on Build GRCh38
rs776575211 Chr.15: 67254880-67254880 on Build GRCh38
rs751036390 Chr.15: 67254853-67254853 on Build GRCh38
rs752798531 Chr.15: 67254977-67254977 on Build GRCh38
rs150409242 Chr.15: 67254888-67254888 on Build GRCh38
rs758889010 Chr.15: 67254907-67254907 on Build GRCh38
rs200974020 Chr.15: 67254948-67254948 on Build GRCh38
rs946493173 Chr.15: 67255014-67255014 on Build GRCh38
rs755165096 Chr.15: 67254922-67254922 on Build GRCh38
rs773018358 Chr.15: 67254884-67254884 on Build GRCh38
rs371158214 Chr.15: 67254957-67254957 on Build GRCh38
rs770780847 Chr.15: 67254936-67254936 on Build GRCh38
rs756208281 Chr.15: 67254982-67254982 on Build GRCh38
rs764498221 Chr.15: 67254856-67254856 on Build GRCh38
rs377745050 Chr.15: 67254924-67254924 on Build GRCh38
rs952758190 Chr.15: 67255049-67255049 on Build GRCh38
rs923304539 Chr.15: 67255018-67255018 on Build GRCh38
rs1027238783 Chr.15: 67254858-67254858 on Build GRCh38
rs749156875 Chr.15: 67254987-67254987 on Build GRCh38
rs1034092213 Chr.15: 67254998-67254998 on Build GRCh38
rs367561206 Chr.15: 67254986-67254986 on Build GRCh38
rs780417908 Chr.15: 67254909-67254909 on Build GRCh38
rs1028398899 Chr.15: 67254946-67254946 on Build GRCh38
rs1028764821 Chr.15: 67255040-67255040 on Build GRCh38
rs145301694 Chr.15: 67254949-67254949 on Build GRCh38
rs746262191 Chr.15: 67254869-67254869 on Build GRCh38
rs140241627 Chr.15: 67254917-67254917 on Build GRCh38
rs755186692 Chr.15: 67254969-67254969 on Build GRCh38
rs1006367107 Chr.15: 67255001-67255001 on Build GRCh38
rs766788620 Chr.15: 67254964-67254964 on Build GRCh38
rs906877522 Chr.15: 67254975-67254975 on Build GRCh38
rs757738757 Chr.15: 67254898-67254898 on Build GRCh38
rs529318414 Chr.15: 67254883-67254883 on Build GRCh38
rs544714275 Chr.15: 67255036-67255036 on Build GRCh38
rs569611185 Chr.15: 67255013-67255013 on Build GRCh38
rs1041308137 Chr.15: 67255024-67255024 on Build GRCh38
rs766986629 Chr.15: 67254890-67254890 on Build GRCh38
rs932015152 Chr.15: 67254891-67254891 on Build GRCh38
rs755260706 Chr.15: 67254968-67254969 on Build GRCh38
rs535472464 Chr.15: 67255050-67255050 on Build GRCh38
rs760146170 Chr.15: 67254956-67254956 on Build GRCh38
rs997305878 Chr.15: 67254925-67254925 on Build GRCh38
rs935833929 Chr.15: 67254943-67254943 on Build GRCh38
rs1043195261 Chr.15: 67255015-67255015 on Build GRCh38

View Full Product Details