Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134991069-134991312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM10
Entrez Gene ID
644538
Gene Name
small integral membrane protein 10
Gene Aliases
CXorf69
UniGene
Hs.42239
Gene Chromosome Location
Chr.X: 134990938-134992473 on Build GRCh38
Transcripts
NM_001163438.1

SNP Detail

SNP ID Chromosome Location
rs759251454 Chr.X: 134991187-134991187 on Build GRCh38
rs775498663 Chr.X: 134991233-134991233 on Build GRCh38
rs777459105 Chr.X: 134991127-134991127 on Build GRCh38
rs768814762 Chr.X: 134991271-134991271 on Build GRCh38
rs1050896508 Chr.X: 134991141-134991141 on Build GRCh38
rs962744897 Chr.X: 134991249-134991249 on Build GRCh38
rs759696194 Chr.X: 134991198-134991198 on Build GRCh38
rs776807747 Chr.X: 134991148-134991148 on Build GRCh38
rs886916848 Chr.X: 134991151-134991151 on Build GRCh38
rs879132744 Chr.X: 134991107-134991107 on Build GRCh38
rs765169017 Chr.X: 134991229-134991229 on Build GRCh38
rs775876425 Chr.X: 134991236-134991236 on Build GRCh38
rs1035571140 Chr.X: 134991116-134991116 on Build GRCh38
rs749634585 Chr.X: 134991241-134991241 on Build GRCh38
rs998046692 Chr.X: 134991123-134991123 on Build GRCh38
rs756721420 Chr.X: 134991202-134991202 on Build GRCh38
rs771193829 Chr.X: 134991103-134991103 on Build GRCh38
rs3180615 Chr.X: 134991113-134991113 on Build GRCh38
rs964697312 Chr.X: 134991221-134991221 on Build GRCh38
rs746568290 Chr.X: 134991201-134991201 on Build GRCh38
rs758163284 Chr.X: 134991091-134991091 on Build GRCh38
rs781564498 Chr.X: 134991213-134991213 on Build GRCh38
rs1047473 Chr.X: 134991179-134991179 on Build GRCh38

View Full Product Details