Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35906131-35906392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRCT1
Entrez Gene ID
646962
Gene Name
histidine rich carboxyl terminus 1
Gene Aliases
LGLL338, PRO537, UNQ338
UniGene
Hs.208081
Gene Chromosome Location
Chr.9: 35906192-35907141 on Build GRCh38
Transcripts
NM_001039792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3375244 c.60G>A p.A20A Chr.9: 35906347-35906347 on Build GRCh38
1490012 c.64_66delCTG p.L28delL Chr.9: 35906351-35906353 on Build GRCh38
5532412 c.103G>A p.G35R Chr.9: 35906390-35906390 on Build GRCh38
1108856 c.46G>A p.G16S Chr.9: 35906333-35906333 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs183527391 Chr.9: 35906373-35906373 on Build GRCh38
rs367568239 Chr.9: 35906254-35906254 on Build GRCh38
rs778003739 Chr.9: 35906264-35906264 on Build GRCh38
rs999034704 Chr.9: 35906338-35906338 on Build GRCh38
rs749449475 Chr.9: 35906271-35906271 on Build GRCh38
rs747464375 Chr.9: 35906279-35906279 on Build GRCh38
rs751344430 Chr.9: 35906342-35906342 on Build GRCh38
rs780289218 Chr.9: 35906222-35906222 on Build GRCh38
rs756325198 Chr.9: 35906262-35906262 on Build GRCh38
rs894542009 Chr.9: 35906277-35906277 on Build GRCh38
rs1036384433 Chr.9: 35906186-35906186 on Build GRCh38
rs530305319 Chr.9: 35906318-35906318 on Build GRCh38
rs150808535 Chr.9: 35906315-35906315 on Build GRCh38
rs771326655 Chr.9: 35906278-35906278 on Build GRCh38
rs944602740 Chr.9: 35906171-35906171 on Build GRCh38
rs561611798 Chr.9: 35906317-35906317 on Build GRCh38
rs770170658 Chr.9: 35906351-35906356 on Build GRCh38
rs374590028 Chr.9: 35906347-35906347 on Build GRCh38
rs903134409 Chr.9: 35906182-35906182 on Build GRCh38
rs772508699 Chr.9: 35906312-35906312 on Build GRCh38
rs955274473 Chr.9: 35906151-35906151 on Build GRCh38
rs375546241 Chr.9: 35906350-35906350 on Build GRCh38
rs796807555 Chr.9: 35906194-35906194 on Build GRCh38
rs777683457 Chr.9: 35906372-35906372 on Build GRCh38
rs370606246 Chr.9: 35906351-35906353 on Build GRCh38
rs1055921741 Chr.9: 35906272-35906272 on Build GRCh38
rs544919035 Chr.9: 35906316-35906316 on Build GRCh38
rs772241532 Chr.9: 35906180-35906180 on Build GRCh38
rs776869376 Chr.9: 35906350-35906351 on Build GRCh38
rs746430989 Chr.9: 35906299-35906299 on Build GRCh38
rs371695108 Chr.9: 35906257-35906257 on Build GRCh38
rs752745134 Chr.9: 35906357-35906357 on Build GRCh38
rs369837960 Chr.9: 35906346-35906346 on Build GRCh38
rs1042023322 Chr.9: 35906178-35906178 on Build GRCh38
rs766340247 Chr.9: 35906339-35906339 on Build GRCh38
rs756267312 Chr.9: 35906367-35906367 on Build GRCh38
rs987032280 Chr.9: 35906170-35906170 on Build GRCh38
rs377191783 Chr.9: 35906283-35906283 on Build GRCh38
rs573234052 Chr.9: 35906330-35906330 on Build GRCh38
rs564612966 Chr.9: 35906152-35906152 on Build GRCh38

View Full Product Details