Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124038042-124038267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10G7
Entrez Gene ID
390265
Gene Name
olfactory receptor family 10 subfamily G member 7
Gene Aliases
OR11-283
UniGene
Hs.553761
Gene Chromosome Location
Chr.11: 124038066-124039001 on Build GRCh38
Transcripts
NM_001004463.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1293330 c.840T>C p.P280P Chr.11: 124038162-124038162 on Build GRCh38
5781704 c.910G>A p.G304R Chr.11: 124038092-124038092 on Build GRCh38
6531332 c.884A>G p.K295R Chr.11: 124038118-124038118 on Build GRCh38
6569034 c.822C>T p.F274F Chr.11: 124038180-124038180 on Build GRCh38
4975003 c.747C>T p.C249C Chr.11: 124038255-124038255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773507107 Chr.11: 124038138-124038138 on Build GRCh38
rs766911838 Chr.11: 124038149-124038149 on Build GRCh38
rs142972044 Chr.11: 124038184-124038185 on Build GRCh38
rs755802892 Chr.11: 124038171-124038171 on Build GRCh38
rs747047250 Chr.11: 124038185-124038185 on Build GRCh38
rs755763606 Chr.11: 124038246-124038246 on Build GRCh38
rs748390521 Chr.11: 124038205-124038205 on Build GRCh38
rs143478590 Chr.11: 124038132-124038132 on Build GRCh38
rs148603378 Chr.11: 124038235-124038235 on Build GRCh38
rs895821309 Chr.11: 124038183-124038183 on Build GRCh38
rs747137233 Chr.11: 124038114-124038114 on Build GRCh38
rs747845511 Chr.11: 124038107-124038107 on Build GRCh38
rs1029651371 Chr.11: 124038103-124038103 on Build GRCh38
rs751136418 Chr.11: 124038077-124038077 on Build GRCh38
rs199934441 Chr.11: 124038209-124038209 on Build GRCh38
rs368332049 Chr.11: 124038164-124038164 on Build GRCh38
rs372451457 Chr.11: 124038223-124038223 on Build GRCh38
rs754592994 Chr.11: 124038087-124038087 on Build GRCh38
rs780871212 Chr.11: 124038100-124038100 on Build GRCh38
rs184333829 Chr.11: 124038176-124038176 on Build GRCh38
rs751044514 Chr.11: 124038152-124038152 on Build GRCh38
rs997119184 Chr.11: 124038099-124038099 on Build GRCh38
rs768779066 Chr.11: 124038242-124038242 on Build GRCh38
rs769977171 Chr.11: 124038206-124038206 on Build GRCh38
rs776944826 Chr.11: 124038125-124038125 on Build GRCh38
rs114086709 Chr.11: 124038172-124038172 on Build GRCh38
rs754504836 Chr.11: 124038153-124038153 on Build GRCh38
rs777501224 Chr.11: 124038109-124038109 on Build GRCh38
rs964616204 Chr.11: 124038078-124038078 on Build GRCh38
rs867513894 Chr.11: 124038234-124038234 on Build GRCh38
rs775683985 Chr.11: 124038112-124038112 on Build GRCh38
rs747090731 Chr.11: 124038175-124038175 on Build GRCh38
rs748463459 Chr.11: 124038194-124038208 on Build GRCh38
rs752203237 Chr.11: 124038240-124038240 on Build GRCh38
rs776659432 Chr.11: 124038199-124038199 on Build GRCh38
rs768954231 Chr.11: 124038120-124038120 on Build GRCh38
rs1044988464 Chr.11: 124038179-124038179 on Build GRCh38
rs765648153 Chr.11: 124038135-124038135 on Build GRCh38
rs774866321 Chr.11: 124038217-124038217 on Build GRCh38
rs947951769 Chr.11: 124038174-124038174 on Build GRCh38
rs528549128 Chr.11: 124038207-124038207 on Build GRCh38
rs767034136 Chr.11: 124038163-124038163 on Build GRCh38
rs927624601 Chr.11: 124038197-124038197 on Build GRCh38
rs749258039 Chr.11: 124038110-124038110 on Build GRCh38
rs763444511 Chr.11: 124038148-124038148 on Build GRCh38
rs553750424 Chr.11: 124038104-124038104 on Build GRCh38

View Full Product Details