Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75555937-75556180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM231
Entrez Gene ID
79583
Gene Name
transmembrane protein 231
Gene Aliases
ALYE870, JBTS20, MKS11, PRO1886
UniGene
Hs.156784
Gene Chromosome Location
Chr.16: 75538117-75556286 on Build GRCh38
Transcripts
NM_001077416.2 NM_001077418.2 NR_074083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1229655 c.151A>G p.K51E Chr.16: 75555962-75555962 on Build GRCh38
1229657 c.164A>C p.Y55S Chr.16: 75555949-75555949 on Build GRCh38
7220390 c.85C>T p.R29C Chr.16: 75556115-75556115 on Build GRCh38
5805743 c.116T>C p.L39P Chr.16: 75556094-75556094 on Build GRCh38
3378049 c.108C>G p.C36W Chr.16: 75556092-75556092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773052053 Chr.16: 75556044-75556044 on Build GRCh38
rs775480392 Chr.16: 75556065-75556065 on Build GRCh38
rs370005084 Chr.16: 75556043-75556043 on Build GRCh38
rs375718217 Chr.16: 75556029-75556029 on Build GRCh38
rs375353411 Chr.16: 75556001-75556001 on Build GRCh38
rs765091642 Chr.16: 75555990-75555990 on Build GRCh38
rs937269706 Chr.16: 75556059-75556059 on Build GRCh38
rs951457866 Chr.16: 75556137-75556137 on Build GRCh38
rs776673877 Chr.16: 75555976-75555976 on Build GRCh38
rs758878104 Chr.16: 75556120-75556120 on Build GRCh38
rs200063331 Chr.16: 75556024-75556024 on Build GRCh38
rs746685376 Chr.16: 75556138-75556138 on Build GRCh38
rs766491956 Chr.16: 75556101-75556101 on Build GRCh38
rs369232571 Chr.16: 75556020-75556020 on Build GRCh38
rs202215735 Chr.16: 75556119-75556119 on Build GRCh38
rs1032603822 Chr.16: 75556037-75556037 on Build GRCh38
rs377416174 Chr.16: 75556126-75556126 on Build GRCh38
rs765968253 Chr.16: 75555955-75555955 on Build GRCh38
rs750621210 Chr.16: 75556008-75556008 on Build GRCh38
rs752366571 Chr.16: 75555991-75555991 on Build GRCh38
rs377440297 Chr.16: 75556081-75556081 on Build GRCh38
rs753458985 Chr.16: 75556098-75556098 on Build GRCh38
rs765152369 Chr.16: 75556064-75556064 on Build GRCh38
rs563689568 Chr.16: 75556012-75556012 on Build GRCh38
rs370607340 Chr.16: 75556131-75556131 on Build GRCh38
rs201518524 Chr.16: 75555988-75555988 on Build GRCh38
rs373488530 Chr.16: 75556071-75556071 on Build GRCh38
rs201181950 Chr.16: 75556115-75556115 on Build GRCh38
rs750674092 Chr.16: 75556080-75556080 on Build GRCh38
rs770598456 Chr.16: 75556048-75556048 on Build GRCh38
rs755383782 Chr.16: 75556018-75556018 on Build GRCh38
rs368402335 Chr.16: 75556079-75556079 on Build GRCh38
rs529302886 Chr.16: 75556051-75556051 on Build GRCh38
rs746632596 Chr.16: 75556047-75556047 on Build GRCh38
rs770637325 Chr.16: 75556159-75556159 on Build GRCh38
rs1012758156 Chr.16: 75556092-75556092 on Build GRCh38
rs901311435 Chr.16: 75556006-75556006 on Build GRCh38
rs755223588 Chr.16: 75556039-75556039 on Build GRCh38
rs971955148 Chr.16: 75556129-75556129 on Build GRCh38
rs759518508 Chr.16: 75556054-75556054 on Build GRCh38
rs767782404 Chr.16: 75556007-75556007 on Build GRCh38
rs751903254 Chr.16: 75555981-75555982 on Build GRCh38
rs780891174 Chr.16: 75556163-75556163 on Build GRCh38
rs756225620 Chr.16: 75556010-75556010 on Build GRCh38
rs749822255 Chr.16: 75556014-75556014 on Build GRCh38
rs753795498 Chr.16: 75556106-75556106 on Build GRCh38

View Full Product Details