Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203056551-203056810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIA4
Entrez Gene ID
8497
Gene Name
PTPRF interacting protein alpha 4
Gene Aliases
-
UniGene
Hs.153648
Gene Chromosome Location
Chr.1: 203026491-203078736 on Build GRCh38
Transcripts
NM_001304331.1 NM_001304332.1 XM_006711586.2 XM_011510070.2 XM_017002606.1 XM_017002607.1 XM_017002608.1 XM_017002609.1 XM_017002610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3723151 c.730G>A p.E244K Chr.1: 203056791-203056791 on Build GRCh38
4788563 c.737A>T p.Q246L Chr.1: 203056798-203056798 on Build GRCh38
6009735 c.723-10C>T p.? Chr.1: 203056774-203056774 on Build GRCh38
3966191 c.749C>G p.P250R Chr.1: 203056810-203056810 on Build GRCh38
1146129 c.748C>A p.P250T Chr.1: 203056809-203056809 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540964051 Chr.1: 203056718-203056718 on Build GRCh38
rs113771133 Chr.1: 203056650-203056650 on Build GRCh38
rs898099579 Chr.1: 203056726-203056726 on Build GRCh38
rs1045049352 Chr.1: 203056593-203056593 on Build GRCh38
rs939570645 Chr.1: 203056676-203056676 on Build GRCh38
rs754847247 Chr.1: 203056734-203056734 on Build GRCh38
rs981359527 Chr.1: 203056791-203056791 on Build GRCh38
rs1026384201 Chr.1: 203056665-203056665 on Build GRCh38
rs984808097 Chr.1: 203056721-203056721 on Build GRCh38
rs746094014 Chr.1: 203056782-203056782 on Build GRCh38
rs572747542 Chr.1: 203056725-203056725 on Build GRCh38
rs747054831 Chr.1: 203056753-203056753 on Build GRCh38
rs372376633 Chr.1: 203056774-203056774 on Build GRCh38
rs1019029300 Chr.1: 203056646-203056646 on Build GRCh38
rs770244758 Chr.1: 203056785-203056785 on Build GRCh38
rs368375485 Chr.1: 203056758-203056758 on Build GRCh38
rs772797942 Chr.1: 203056652-203056652 on Build GRCh38
rs879154074 Chr.1: 203056717-203056717 on Build GRCh38
rs778949920 Chr.1: 203056741-203056741 on Build GRCh38
rs1056735995 Chr.1: 203056716-203056716 on Build GRCh38
rs61217599 Chr.1: 203056757-203056757 on Build GRCh38
rs952252840 Chr.1: 203056707-203056707 on Build GRCh38
rs770278630 Chr.1: 203056748-203056748 on Build GRCh38
rs993512859 Chr.1: 203056659-203056659 on Build GRCh38
rs757862504 Chr.1: 203056775-203056775 on Build GRCh38
rs781728688 Chr.1: 203056777-203056777 on Build GRCh38
rs889366994 Chr.1: 203056588-203056588 on Build GRCh38
rs1007772815 Chr.1: 203056645-203056645 on Build GRCh38

View Full Product Details