Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166405710-166405951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN7A
Entrez Gene ID
6332
Gene Name
sodium voltage-gated channel alpha subunit 7
Gene Aliases
NaG, Nav2.1, Nav2.2, SCN6A
UniGene
Hs.596087
Gene Chromosome Location
Chr.2: 166403573-166494264 on Build GRCh38
Transcripts
NM_002976.3 NR_045628.1 XM_006712680.2 XM_006712681.3 XM_006712682.3 XM_011511615.2 XM_017004667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
126044 c.4900C>A p.R1634S Chr.2: 166405729-166405729 on Build GRCh38
4417347 c.4786G>C p.V1596L Chr.2: 166405843-166405843 on Build GRCh38
3569281 c.4865G>A p.R1622Q Chr.2: 166405764-166405764 on Build GRCh38
6904858 c.4835C>T p.T1612I Chr.2: 166405794-166405794 on Build GRCh38
2153491 c.4815A>G p.L1605L Chr.2: 166405814-166405814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757402063 Chr.2: 166405904-166405904 on Build GRCh38
rs760432847 Chr.2: 166405894-166405895 on Build GRCh38
rs781244691 Chr.2: 166405898-166405898 on Build GRCh38
rs533803665 Chr.2: 166405807-166405807 on Build GRCh38
rs1039617808 Chr.2: 166405816-166405816 on Build GRCh38
rs772318275 Chr.2: 166405793-166405793 on Build GRCh38
rs994942330 Chr.2: 166405838-166405838 on Build GRCh38
rs765043857 Chr.2: 166405740-166405740 on Build GRCh38
rs775668770 Chr.2: 166405930-166405930 on Build GRCh38
rs912169071 Chr.2: 166405782-166405782 on Build GRCh38
rs571278405 Chr.2: 166405765-166405765 on Build GRCh38
rs772498030 Chr.2: 166405927-166405927 on Build GRCh38
rs367571939 Chr.2: 166405832-166405832 on Build GRCh38
rs764916096 Chr.2: 166405846-166405846 on Build GRCh38
rs767479047 Chr.2: 166405826-166405826 on Build GRCh38
rs750060751 Chr.2: 166405847-166405847 on Build GRCh38
rs899220196 Chr.2: 166405836-166405836 on Build GRCh38
rs77304082 Chr.2: 166405920-166405920 on Build GRCh38
rs766032987 Chr.2: 166405872-166405872 on Build GRCh38
rs771319659 Chr.2: 166405778-166405778 on Build GRCh38
rs776076462 Chr.2: 166405834-166405834 on Build GRCh38
rs373935271 Chr.2: 166405861-166405861 on Build GRCh38
rs780094202 Chr.2: 166405754-166405754 on Build GRCh38
rs759394259 Chr.2: 166405825-166405826 on Build GRCh38
rs202047112 Chr.2: 166405894-166405894 on Build GRCh38
rs764963248 Chr.2: 166405758-166405758 on Build GRCh38
rs3791251 Chr.2: 166405843-166405843 on Build GRCh38
rs746294875 Chr.2: 166405787-166405787 on Build GRCh38
rs776067124 Chr.2: 166405806-166405806 on Build GRCh38
rs372024247 Chr.2: 166405903-166405903 on Build GRCh38
rs773283261 Chr.2: 166405775-166405775 on Build GRCh38
rs750254009 Chr.2: 166405750-166405750 on Build GRCh38
rs752539431 Chr.2: 166405834-166405834 on Build GRCh38
rs749554142 Chr.2: 166405768-166405768 on Build GRCh38
rs779346962 Chr.2: 166405910-166405910 on Build GRCh38
rs746020061 Chr.2: 166405926-166405926 on Build GRCh38
rs751541619 Chr.2: 166405877-166405877 on Build GRCh38
rs374349539 Chr.2: 166405779-166405779 on Build GRCh38
rs758164669 Chr.2: 166405752-166405752 on Build GRCh38
rs188781935 Chr.2: 166405764-166405764 on Build GRCh38

View Full Product Details