Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 87836312-87836537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A40
Entrez Gene ID
55972
Gene Name
solute carrier family 25 member 40
Gene Aliases
MCFP
UniGene
Hs.208414
Gene Chromosome Location
Chr.7: 87834499-87876377 on Build GRCh38
Transcripts
NM_018843.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1092661 c.940T>G p.C314G Chr.7: 87836326-87836326 on Build GRCh38
5962521 c.922A>G p.I308V Chr.7: 87836344-87836344 on Build GRCh38
5737984 c.932C>T p.A311V Chr.7: 87836334-87836334 on Build GRCh38
747718 c.905G>C p.G302A Chr.7: 87836361-87836361 on Build GRCh38
6012768 c.913C>T p.P305S Chr.7: 87836353-87836353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779693303 Chr.7: 87836389-87836389 on Build GRCh38
rs752371624 Chr.7: 87836445-87836445 on Build GRCh38
rs760581054 Chr.7: 87836394-87836394 on Build GRCh38
rs531765370 Chr.7: 87836416-87836416 on Build GRCh38
rs984570654 Chr.7: 87836434-87836434 on Build GRCh38
rs763297221 Chr.7: 87836365-87836369 on Build GRCh38
rs774650967 Chr.7: 87836393-87836395 on Build GRCh38
rs950794753 Chr.7: 87836468-87836468 on Build GRCh38
rs764240725 Chr.7: 87836381-87836381 on Build GRCh38
rs775429538 Chr.7: 87836349-87836349 on Build GRCh38
rs973817328 Chr.7: 87836427-87836427 on Build GRCh38
rs373485583 Chr.7: 87836376-87836376 on Build GRCh38
rs371373274 Chr.7: 87836423-87836423 on Build GRCh38
rs545972367 Chr.7: 87836357-87836357 on Build GRCh38
rs1026368081 Chr.7: 87836459-87836459 on Build GRCh38
rs759663615 Chr.7: 87836350-87836350 on Build GRCh38
rs987291585 Chr.7: 87836487-87836487 on Build GRCh38
rs951896191 Chr.7: 87836425-87836425 on Build GRCh38
rs753835874 Chr.7: 87836360-87836360 on Build GRCh38
rs763379005 Chr.7: 87836347-87836347 on Build GRCh38
rs369225044 Chr.7: 87836373-87836373 on Build GRCh38
rs201654319 Chr.7: 87836374-87836374 on Build GRCh38
rs746578670 Chr.7: 87836395-87836395 on Build GRCh38
rs766776279 Chr.7: 87836349-87836349 on Build GRCh38
rs766223845 Chr.7: 87836395-87836395 on Build GRCh38

View Full Product Details