Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19296924-19297151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUGP1
Entrez Gene ID
57794
Gene Name
SURP and G-patch domain containing 1
Gene Aliases
F23858, RBP, SF4
UniGene
Hs.515274
Gene Chromosome Location
Chr.19: 19276513-19320512 on Build GRCh38
Transcripts
NM_172231.3 XM_005260002.2 XM_011528155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7242793 c.1099C>G p.P367A Chr.19: 19297133-19297133 on Build GRCh38
5962333 c.1184A>G p.E395G Chr.19: 19297048-19297048 on Build GRCh38
340497 c.1157G>T p.R386L Chr.19: 19297075-19297075 on Build GRCh38
1744483 c.1243G>T p.V415F Chr.19: 19296989-19296989 on Build GRCh38
5661522 c.1216G>T p.V406L Chr.19: 19297016-19297016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141492378 Chr.19: 19297107-19297107 on Build GRCh38
rs1038134336 Chr.19: 19296982-19296982 on Build GRCh38
rs143869423 Chr.19: 19296971-19296971 on Build GRCh38
rs934156139 Chr.19: 19297014-19297014 on Build GRCh38
rs779289393 Chr.19: 19296999-19296999 on Build GRCh38
rs767598998 Chr.19: 19297120-19297120 on Build GRCh38
rs765296196 Chr.19: 19297030-19297030 on Build GRCh38
rs1013287126 Chr.19: 19297010-19297010 on Build GRCh38
rs771173241 Chr.19: 19297020-19297020 on Build GRCh38
rs750287204 Chr.19: 19297055-19297055 on Build GRCh38
rs200062643 Chr.19: 19297106-19297106 on Build GRCh38
rs922806185 Chr.19: 19297005-19297005 on Build GRCh38
rs143568455 Chr.19: 19297078-19297078 on Build GRCh38
rs761900209 Chr.19: 19297043-19297043 on Build GRCh38
rs764761779 Chr.19: 19297131-19297131 on Build GRCh38
rs554234174 Chr.19: 19296969-19296969 on Build GRCh38
rs747395123 Chr.19: 19297097-19297097 on Build GRCh38
rs773612129 Chr.19: 19297012-19297012 on Build GRCh38
rs866478381 Chr.19: 19297076-19297076 on Build GRCh38
rs767543906 Chr.19: 19297044-19297044 on Build GRCh38
rs748782198 Chr.19: 19296991-19296991 on Build GRCh38
rs755934845 Chr.19: 19297068-19297068 on Build GRCh38
rs758942961 Chr.19: 19297085-19297085 on Build GRCh38
rs772549956 Chr.19: 19297006-19297006 on Build GRCh38
rs373152885 Chr.19: 19297124-19297124 on Build GRCh38
rs759618518 Chr.19: 19296966-19296966 on Build GRCh38
rs998515034 Chr.19: 19297084-19297084 on Build GRCh38
rs545601901 Chr.19: 19297122-19297122 on Build GRCh38
rs865923116 Chr.19: 19297127-19297127 on Build GRCh38
rs773238000 Chr.19: 19297121-19297121 on Build GRCh38
rs757289120 Chr.19: 19296978-19296978 on Build GRCh38
rs778212982 Chr.19: 19297094-19297094 on Build GRCh38
rs115426450 Chr.19: 19297074-19297074 on Build GRCh38
rs757124605 Chr.19: 19296970-19296970 on Build GRCh38
rs111849423 Chr.19: 19297017-19297017 on Build GRCh38
rs761145320 Chr.19: 19296952-19296952 on Build GRCh38
rs747630971 Chr.19: 19297083-19297083 on Build GRCh38
rs767527075 Chr.19: 19297024-19297024 on Build GRCh38
rs984873300 Chr.19: 19296952-19296952 on Build GRCh38
rs148031252 Chr.19: 19297098-19297098 on Build GRCh38
rs754579542 Chr.19: 19297079-19297079 on Build GRCh38
rs778539130 Chr.19: 19297080-19297080 on Build GRCh38
rs908608786 Chr.19: 19296967-19296967 on Build GRCh38
rs370183395 Chr.19: 19296975-19296975 on Build GRCh38
rs768166418 Chr.19: 19296993-19296993 on Build GRCh38
rs759549480 Chr.19: 19297029-19297029 on Build GRCh38
rs1049880786 Chr.19: 19297016-19297016 on Build GRCh38
rs376087890 Chr.19: 19297126-19297126 on Build GRCh38
rs763097381 Chr.19: 19297111-19297111 on Build GRCh38
rs764210227 Chr.19: 19296964-19296964 on Build GRCh38
rs775688929 Chr.19: 19297109-19297109 on Build GRCh38
rs775427869 Chr.19: 19297031-19297031 on Build GRCh38

View Full Product Details