Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18636415-18636652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D28
Entrez Gene ID
254272
Gene Name
TBC1 domain family member 28
Gene Aliases
-
UniGene
Hs.434123
Gene Chromosome Location
Chr.17: 18634434-18644431 on Build GRCh38
Transcripts
NM_001039397.2 XM_011523781.2 XM_011523782.2 XM_011523783.2 XM_017024416.1 XM_017024417.1 XM_017024418.1 XM_017024419.1 XM_017024420.1 XM_017024421.1 XM_017024422.1 XM_017024423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6016532 c.626C>G p.T209S Chr.17: 18636469-18636469 on Build GRCh38
300344 c.559G>A p.G187R Chr.17: 18636536-18636536 on Build GRCh38
2741187 c.617G>A p.G206D Chr.17: 18636478-18636478 on Build GRCh38
5754992 c.581T>C p.L194P Chr.17: 18636514-18636514 on Build GRCh38
4567080 c.556_557CC>TT p.P186F Chr.17: 18636538-18636539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375442485 Chr.17: 18636576-18636576 on Build GRCh38
rs770092316 Chr.17: 18636505-18636505 on Build GRCh38
rs752722188 Chr.17: 18636532-18636532 on Build GRCh38
rs756076458 Chr.17: 18636534-18636534 on Build GRCh38
rs763855902 Chr.17: 18636536-18636536 on Build GRCh38
rs767683127 Chr.17: 18636530-18636530 on Build GRCh38
rs766631113 Chr.17: 18636519-18636519 on Build GRCh38
rs748667658 Chr.17: 18636561-18636561 on Build GRCh38
rs751794727 Chr.17: 18636521-18636521 on Build GRCh38
rs764994643 Chr.17: 18636623-18636623 on Build GRCh38
rs757094365 Chr.17: 18636608-18636608 on Build GRCh38
rs766433599 Chr.17: 18636463-18636463 on Build GRCh38
rs775700158 Chr.17: 18636593-18636593 on Build GRCh38
rs755045064 Chr.17: 18636468-18636468 on Build GRCh38
rs762685813 Chr.17: 18636599-18636599 on Build GRCh38
rs748948782 Chr.17: 18636439-18636439 on Build GRCh38
rs778478691 Chr.17: 18636484-18636484 on Build GRCh38
rs745474005 Chr.17: 18636549-18636549 on Build GRCh38
rs761226219 Chr.17: 18636447-18636447 on Build GRCh38
rs769595278 Chr.17: 18636436-18636436 on Build GRCh38
rs777141368 Chr.17: 18636498-18636498 on Build GRCh38
rs537403850 Chr.17: 18636465-18636465 on Build GRCh38
rs745385564 Chr.17: 18636487-18636487 on Build GRCh38
rs760612092 Chr.17: 18636595-18636595 on Build GRCh38
rs369754301 Chr.17: 18636537-18636537 on Build GRCh38
rs369463459 Chr.17: 18636590-18636590 on Build GRCh38
rs201749333 Chr.17: 18636624-18636625 on Build GRCh38
rs368055633 Chr.17: 18636469-18636469 on Build GRCh38
rs397834032 Chr.17: 18636515-18636515 on Build GRCh38
rs397834033 Chr.17: 18636518-18636518 on Build GRCh38
rs375857329 Chr.17: 18636482-18636482 on Build GRCh38
rs758040024 Chr.17: 18636630-18636630 on Build GRCh38
rs375356825 Chr.17: 18636500-18636500 on Build GRCh38
rs372335213 Chr.17: 18636491-18636491 on Build GRCh38
rs557338602 Chr.17: 18636624-18636624 on Build GRCh38
rs965015774 Chr.17: 18636570-18636570 on Build GRCh38
rs370901643 Chr.17: 18636437-18636437 on Build GRCh38
rs759715864 Chr.17: 18636523-18636523 on Build GRCh38
rs1031818240 Chr.17: 18636516-18636516 on Build GRCh38
rs756933847 Chr.17: 18636541-18636541 on Build GRCh38
rs535416346 Chr.17: 18636568-18636568 on Build GRCh38
rs777055677 Chr.17: 18636451-18636451 on Build GRCh38
rs375540219 Chr.17: 18636452-18636452 on Build GRCh38
rs766127769 Chr.17: 18636619-18636620 on Build GRCh38
rs750620319 Chr.17: 18636552-18636558 on Build GRCh38
rs779595061 Chr.17: 18636631-18636631 on Build GRCh38
rs753674568 Chr.17: 18636606-18636606 on Build GRCh38
rs764094277 Chr.17: 18636598-18636598 on Build GRCh38
rs773909957 Chr.17: 18636445-18636445 on Build GRCh38
rs774763431 Chr.17: 18636574-18636574 on Build GRCh38
rs779504959 Chr.17: 18636558-18636558 on Build GRCh38
rs770602004 Chr.17: 18636483-18636483 on Build GRCh38
rs757948094 Chr.17: 18636557-18636557 on Build GRCh38
rs770388828 Chr.17: 18636440-18636440 on Build GRCh38
rs763200541 Chr.17: 18636453-18636453 on Build GRCh38
rs373183034 Chr.17: 18636573-18636573 on Build GRCh38
rs371584400 Chr.17: 18636481-18636481 on Build GRCh38
rs202167244 Chr.17: 18636572-18636572 on Build GRCh38
rs778566943 Chr.17: 18636543-18636543 on Build GRCh38

View Full Product Details