Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132294560-132294821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A4
Entrez Gene ID
6583
Gene Name
solute carrier family 22 member 4
Gene Aliases
OCTN1
UniGene
Hs.310591
Gene Chromosome Location
Chr.5: 132294384-132344206 on Build GRCh38
Transcripts
NM_003059.2 XM_011543589.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6168681 c.167C>G p.A56G Chr.5: 132294783-132294783 on Build GRCh38
6168682 c.82A>G p.S28G Chr.5: 132294698-132294698 on Build GRCh38
1495915 c.4C>T p.R2W Chr.5: 132294620-132294620 on Build GRCh38
7197821 c.62T>A p.I21N Chr.5: 132294678-132294678 on Build GRCh38
6455844 c.95A>G p.N32S Chr.5: 132294711-132294711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750134122 Chr.5: 132294757-132294757 on Build GRCh38
rs762771154 Chr.5: 132294722-132294722 on Build GRCh38
rs565099252 Chr.5: 132294697-132294697 on Build GRCh38
rs751103752 Chr.5: 132294641-132294641 on Build GRCh38
rs531041134 Chr.5: 132294774-132294774 on Build GRCh38
rs11568501 Chr.5: 132294607-132294607 on Build GRCh38
rs936998472 Chr.5: 132294744-132294744 on Build GRCh38
rs971116318 Chr.5: 132294691-132294691 on Build GRCh38
rs761375217 Chr.5: 132294753-132294753 on Build GRCh38
rs780840704 Chr.5: 132294653-132294653 on Build GRCh38
rs764024260 Chr.5: 132294736-132294736 on Build GRCh38
rs766127231 Chr.5: 132294803-132294807 on Build GRCh38
rs750031077 Chr.5: 132294667-132294667 on Build GRCh38
rs764924955 Chr.5: 132294625-132294625 on Build GRCh38
rs774216692 Chr.5: 132294750-132294750 on Build GRCh38
rs747505093 Chr.5: 132294782-132294782 on Build GRCh38
rs191877372 Chr.5: 132294592-132294592 on Build GRCh38
rs914846394 Chr.5: 132294759-132294759 on Build GRCh38
rs777050701 Chr.5: 132294706-132294706 on Build GRCh38
rs772836216 Chr.5: 132294749-132294749 on Build GRCh38
rs774010784 Chr.5: 132294637-132294637 on Build GRCh38
rs372949734 Chr.5: 132294711-132294711 on Build GRCh38
rs778384141 Chr.5: 132294692-132294692 on Build GRCh38
rs567991307 Chr.5: 132294804-132294804 on Build GRCh38
rs775173540 Chr.5: 132294720-132294720 on Build GRCh38
rs909118677 Chr.5: 132294588-132294588 on Build GRCh38
rs771880004 Chr.5: 132294678-132294680 on Build GRCh38
rs752743549 Chr.5: 132294633-132294633 on Build GRCh38
rs760498727 Chr.5: 132294789-132294790 on Build GRCh38
rs762765767 Chr.5: 132294637-132294637 on Build GRCh38
rs756693505 Chr.5: 132294644-132294644 on Build GRCh38
rs747735758 Chr.5: 132294641-132294660 on Build GRCh38
rs189072785 Chr.5: 132294797-132294797 on Build GRCh38
rs926936998 Chr.5: 132294723-132294723 on Build GRCh38
rs199926342 Chr.5: 132294598-132294598 on Build GRCh38
rs375954722 Chr.5: 132294775-132294775 on Build GRCh38
rs759385265 Chr.5: 132294621-132294621 on Build GRCh38
rs760787423 Chr.5: 132294606-132294606 on Build GRCh38
rs889150081 Chr.5: 132294597-132294597 on Build GRCh38
rs747086200 Chr.5: 132294694-132294694 on Build GRCh38
rs778433494 Chr.5: 132294781-132294781 on Build GRCh38
rs962839397 Chr.5: 132294616-132294616 on Build GRCh38
rs763973178 Chr.5: 132294639-132294639 on Build GRCh38
rs767014071 Chr.5: 132294754-132294754 on Build GRCh38
rs545500558 Chr.5: 132294688-132294688 on Build GRCh38
rs758995324 Chr.5: 132294690-132294690 on Build GRCh38
rs755438066 Chr.5: 132294671-132294671 on Build GRCh38
rs779117302 Chr.5: 132294672-132294672 on Build GRCh38
rs776926210 Chr.5: 132294619-132294619 on Build GRCh38
rs781099471 Chr.5: 132294803-132294803 on Build GRCh38
rs1001700978 Chr.5: 132294747-132294747 on Build GRCh38
rs769668837 Chr.5: 132294715-132294715 on Build GRCh38

View Full Product Details