Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74632462-74632735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFWD3
Entrez Gene ID
55159
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
RNF201
UniGene
Hs.567525
Gene Chromosome Location
Chr.16: 74621396-74666892 on Build GRCh38
Transcripts
NM_018124.3 XM_005256021.4 XM_005256022.4 XM_006721228.3 XM_011523191.2 XM_017023391.1 XM_017023392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765340 c.1427-2A>G p.? Chr.16: 74632675-74632675 on Build GRCh38
5851475 c.1451C>T p.T484I Chr.16: 74632649-74632649 on Build GRCh38
704147 c.1457A>G p.N486S Chr.16: 74632643-74632643 on Build GRCh38
973639 c.1441A>G p.M481V Chr.16: 74632659-74632659 on Build GRCh38
3731804 c.1512G>A p.A504A Chr.16: 74632588-74632588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764618501 Chr.16: 74632539-74632539 on Build GRCh38
rs936212251 Chr.16: 74632605-74632605 on Build GRCh38
rs772992025 Chr.16: 74632555-74632556 on Build GRCh38
rs748102081 Chr.16: 74632613-74632613 on Build GRCh38
rs755848003 Chr.16: 74632582-74632582 on Build GRCh38
rs769224672 Chr.16: 74632490-74632490 on Build GRCh38
rs778635619 Chr.16: 74632681-74632681 on Build GRCh38
rs767869165 Chr.16: 74632549-74632549 on Build GRCh38
rs748083506 Chr.16: 74632683-74632683 on Build GRCh38
rs775719646 Chr.16: 74632706-74632706 on Build GRCh38
rs753446117 Chr.16: 74632677-74632677 on Build GRCh38
rs774770054 Chr.16: 74632634-74632634 on Build GRCh38
rs552108764 Chr.16: 74632689-74632689 on Build GRCh38
rs922333404 Chr.16: 74632570-74632570 on Build GRCh38
rs187252203 Chr.16: 74632506-74632506 on Build GRCh38
rs1016851090 Chr.16: 74632708-74632708 on Build GRCh38
rs750829372 Chr.16: 74632647-74632647 on Build GRCh38
rs763035630 Chr.16: 74632627-74632627 on Build GRCh38
rs202130447 Chr.16: 74632641-74632641 on Build GRCh38
rs759902600 Chr.16: 74632523-74632523 on Build GRCh38
rs776928078 Chr.16: 74632519-74632519 on Build GRCh38
rs371120350 Chr.16: 74632491-74632491 on Build GRCh38
rs199852515 Chr.16: 74632619-74632619 on Build GRCh38
rs779404005 Chr.16: 74632595-74632595 on Build GRCh38
rs758102530 Chr.16: 74632603-74632604 on Build GRCh38
rs377365468 Chr.16: 74632659-74632659 on Build GRCh38
rs373095908 Chr.16: 74632707-74632707 on Build GRCh38
rs770650354 Chr.16: 74632699-74632699 on Build GRCh38
rs780771060 Chr.16: 74632488-74632488 on Build GRCh38
rs779676190 Chr.16: 74632492-74632492 on Build GRCh38
rs775594158 Chr.16: 74632677-74632677 on Build GRCh38
rs1045819269 Chr.16: 74632676-74632676 on Build GRCh38
rs769001463 Chr.16: 74632618-74632618 on Build GRCh38
rs757557494 Chr.16: 74632548-74632548 on Build GRCh38
rs752079432 Chr.16: 74632542-74632542 on Build GRCh38
rs895248099 Chr.16: 74632594-74632594 on Build GRCh38
rs897274964 Chr.16: 74632657-74632657 on Build GRCh38
rs769462918 Chr.16: 74632507-74632508 on Build GRCh38
rs772286324 Chr.16: 74632647-74632656 on Build GRCh38
rs752181138 Chr.16: 74632528-74632528 on Build GRCh38
rs759707009 Chr.16: 74632595-74632596 on Build GRCh38
rs374606493 Chr.16: 74632650-74632650 on Build GRCh38
rs887032172 Chr.16: 74632691-74632691 on Build GRCh38
rs758457947 Chr.16: 74632688-74632688 on Build GRCh38
rs778938061 Chr.16: 74632612-74632612 on Build GRCh38
rs748795600 Chr.16: 74632599-74632599 on Build GRCh38
rs761652477 Chr.16: 74632509-74632509 on Build GRCh38
rs754469109 Chr.16: 74632600-74632600 on Build GRCh38
rs764091102 Chr.16: 74632633-74632633 on Build GRCh38
rs750065808 Chr.16: 74632580-74632580 on Build GRCh38
rs377155984 Chr.16: 74632498-74632498 on Build GRCh38
rs768683544 Chr.16: 74632501-74632501 on Build GRCh38
rs770094777 Chr.16: 74632623-74632623 on Build GRCh38
rs200891821 Chr.16: 74632489-74632489 on Build GRCh38
rs963701946 Chr.16: 74632505-74632505 on Build GRCh38
rs756470042 Chr.16: 74632658-74632658 on Build GRCh38
rs1055119135 Chr.16: 74632545-74632545 on Build GRCh38
rs373021544 Chr.16: 74632487-74632487 on Build GRCh38
rs369886677 Chr.16: 74632525-74632525 on Build GRCh38
rs765216000 Chr.16: 74632524-74632524 on Build GRCh38
rs548123185 Chr.16: 74632617-74632617 on Build GRCh38
rs754446035 Chr.16: 74632679-74632679 on Build GRCh38
rs955547385 Chr.16: 74632589-74632589 on Build GRCh38

View Full Product Details