Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73573259-73573524 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBL2
Entrez Gene ID
26608
Gene Name
transducin (beta)-like 2
Gene Aliases
WBSCR13, WS-betaTRP
UniGene
Hs.647044
Gene Chromosome Location
Chr.7: 73568944-73578683 on Build GRCh38
Transcripts
NM_012453.2 XM_006715923.3 XM_011516027.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4960537 c.598+1G>A p.? Chr.7: 73573319-73573319 on Build GRCh38
7187526 c.504G>A p.K168K Chr.7: 73573414-73573414 on Build GRCh38
6797792 c.492C>A p.F164L Chr.7: 73573426-73573426 on Build GRCh38
4499745 c.549C>T p.F183F Chr.7: 73573369-73573369 on Build GRCh38
4497624 c.500C>T p.T167I Chr.7: 73573418-73573418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368230823 Chr.7: 73573413-73573413 on Build GRCh38
rs141209031 Chr.7: 73573351-73573351 on Build GRCh38
rs782117770 Chr.7: 73573313-73573313 on Build GRCh38
rs374299938 Chr.7: 73573371-73573371 on Build GRCh38
rs782307846 Chr.7: 73573395-73573395 on Build GRCh38
rs201440606 Chr.7: 73573464-73573464 on Build GRCh38
rs782741857 Chr.7: 73573434-73573434 on Build GRCh38
rs199565638 Chr.7: 73573341-73573341 on Build GRCh38
rs782343974 Chr.7: 73573474-73573474 on Build GRCh38
rs782266230 Chr.7: 73573471-73573471 on Build GRCh38
rs781961946 Chr.7: 73573297-73573297 on Build GRCh38
rs782631043 Chr.7: 73573475-73573475 on Build GRCh38
rs140134641 Chr.7: 73573462-73573462 on Build GRCh38
rs781929743 Chr.7: 73573396-73573396 on Build GRCh38
rs782365455 Chr.7: 73573296-73573296 on Build GRCh38
rs782041262 Chr.7: 73573312-73573312 on Build GRCh38
rs782494800 Chr.7: 73573447-73573447 on Build GRCh38
rs537053937 Chr.7: 73573407-73573407 on Build GRCh38
rs909772870 Chr.7: 73573404-73573404 on Build GRCh38
rs782074371 Chr.7: 73573298-73573298 on Build GRCh38
rs782068911 Chr.7: 73573441-73573441 on Build GRCh38
rs782796582 Chr.7: 73573443-73573443 on Build GRCh38
rs782704128 Chr.7: 73573348-73573348 on Build GRCh38
rs373168324 Chr.7: 73573461-73573461 on Build GRCh38
rs369202788 Chr.7: 73573497-73573497 on Build GRCh38
rs553085433 Chr.7: 73573489-73573489 on Build GRCh38
rs782018334 Chr.7: 73573491-73573491 on Build GRCh38
rs782252075 Chr.7: 73573292-73573292 on Build GRCh38
rs782673735 Chr.7: 73573352-73573352 on Build GRCh38
rs782798087 Chr.7: 73573335-73573335 on Build GRCh38
rs782329905 Chr.7: 73573490-73573490 on Build GRCh38
rs782596774 Chr.7: 73573364-73573364 on Build GRCh38
rs782698196 Chr.7: 73573309-73573309 on Build GRCh38
rs181232035 Chr.7: 73573345-73573345 on Build GRCh38
rs782002015 Chr.7: 73573291-73573291 on Build GRCh38
rs782789926 Chr.7: 73573448-73573448 on Build GRCh38
rs782240994 Chr.7: 73573355-73573355 on Build GRCh38
rs377105109 Chr.7: 73573498-73573498 on Build GRCh38
rs150732801 Chr.7: 73573460-73573460 on Build GRCh38
rs372356020 Chr.7: 73573433-73573433 on Build GRCh38
rs782509749 Chr.7: 73573359-73573359 on Build GRCh38
rs781831446 Chr.7: 73573342-73573342 on Build GRCh38
rs200461366 Chr.7: 73573305-73573305 on Build GRCh38
rs781926043 Chr.7: 73573439-73573439 on Build GRCh38
rs375440560 Chr.7: 73573446-73573446 on Build GRCh38
rs558316664 Chr.7: 73573412-73573412 on Build GRCh38
rs976555785 Chr.7: 73573299-73573299 on Build GRCh38

View Full Product Details