Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201475548-201475807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRADB
Entrez Gene ID
55437
Gene Name
STE20-related kinase adaptor beta
Gene Aliases
ALS2CR2, CALS-21, ILPIP, ILPIPA, PAPK, PRO1038
UniGene
Hs.652338
Gene Chromosome Location
Chr.2: 201451669-201480851 on Build GRCh38
Transcripts
NM_001206864.1 NM_018571.5 XM_005246669.1 XM_017004438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3933508 c.436C>G p.Q146E Chr.2: 201475630-201475630 on Build GRCh38
6792367 c.451T>C p.Y151H Chr.2: 201475645-201475645 on Build GRCh38
5908165 c.463G>A p.G155R Chr.2: 201475657-201475657 on Build GRCh38
3364495 c.530A>T p.Q177L Chr.2: 201475724-201475724 on Build GRCh38
4341961 c.433A>C p.S145R Chr.2: 201475627-201475627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979048813 Chr.2: 201475700-201475700 on Build GRCh38
rs865949332 Chr.2: 201475738-201475738 on Build GRCh38
rs756037325 Chr.2: 201475681-201475681 on Build GRCh38
rs61742622 Chr.2: 201475734-201475734 on Build GRCh38
rs757539319 Chr.2: 201475579-201475579 on Build GRCh38
rs763789726 Chr.2: 201475628-201475628 on Build GRCh38
rs199922310 Chr.2: 201475717-201475717 on Build GRCh38
rs768465769 Chr.2: 201475698-201475698 on Build GRCh38
rs745424750 Chr.2: 201475646-201475646 on Build GRCh38
rs748372507 Chr.2: 201475716-201475716 on Build GRCh38
rs377363227 Chr.2: 201475699-201475699 on Build GRCh38
rs767051509 Chr.2: 201475592-201475592 on Build GRCh38
rs200634284 Chr.2: 201475595-201475595 on Build GRCh38
rs377322850 Chr.2: 201475762-201475762 on Build GRCh38
rs755406826 Chr.2: 201475619-201475619 on Build GRCh38
rs202000881 Chr.2: 201475692-201475692 on Build GRCh38
rs761243658 Chr.2: 201475585-201475585 on Build GRCh38
rs780727959 Chr.2: 201475644-201475644 on Build GRCh38
rs536861912 Chr.2: 201475647-201475647 on Build GRCh38
rs962930228 Chr.2: 201475601-201475601 on Build GRCh38
rs766934900 Chr.2: 201475766-201475766 on Build GRCh38
rs751255210 Chr.2: 201475639-201475639 on Build GRCh38
rs573046235 Chr.2: 201475754-201475754 on Build GRCh38
rs756898630 Chr.2: 201475640-201475640 on Build GRCh38
rs779975650 Chr.2: 201475684-201475684 on Build GRCh38
rs777299833 Chr.2: 201475777-201475777 on Build GRCh38

View Full Product Details