Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66501280-66501539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL4
Entrez Gene ID
6124
Gene Name
ribosomal protein L4
Gene Aliases
L4
UniGene
Hs.186350
Gene Chromosome Location
Chr.15: 66499315-66504855 on Build GRCh38
Transcripts
NM_000968.3 NR_002443.1

Gene Symbol
SNORD18C
Entrez Gene ID
595100
Gene Name
small nucleolar RNA, C/D box 18C
Gene Aliases
U18C
UniGene
Gene Chromosome Location
Chr.15: 66501252-66501318 on Build GRCh38
Transcripts
NM_000968.3 NR_002443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
416853 c.654C>G p.I218M Chr.15: 66501397-66501397 on Build GRCh38
6398489 c.652A>T p.I218F Chr.15: 66501399-66501399 on Build GRCh38
4955648 c.644A>G p.N215S Chr.15: 66501407-66501407 on Build GRCh38
6397805 c.593A>G p.N198S Chr.15: 66501458-66501458 on Build GRCh38
3503337 c.555C>T p.A185A Chr.15: 66501496-66501496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568596736 Chr.15: 66501431-66501431 on Build GRCh38
rs770220748 Chr.15: 66501314-66501314 on Build GRCh38
rs773116067 Chr.15: 66501345-66501345 on Build GRCh38
rs755648741 Chr.15: 66501312-66501312 on Build GRCh38
rs774269880 Chr.15: 66501439-66501439 on Build GRCh38
rs532024567 Chr.15: 66501368-66501368 on Build GRCh38
rs766975969 Chr.15: 66501414-66501414 on Build GRCh38
rs1012515376 Chr.15: 66501362-66501362 on Build GRCh38
rs146070182 Chr.15: 66501457-66501457 on Build GRCh38
rs11549599 Chr.15: 66501407-66501407 on Build GRCh38
rs11549583 Chr.15: 66501469-66501469 on Build GRCh38
rs993802695 Chr.15: 66501310-66501310 on Build GRCh38
rs374397558 Chr.15: 66501393-66501393 on Build GRCh38
rs749401717 Chr.15: 66501347-66501347 on Build GRCh38
rs761401663 Chr.15: 66501496-66501496 on Build GRCh38
rs755353155 Chr.15: 66501333-66501333 on Build GRCh38
rs139277298 Chr.15: 66501430-66501430 on Build GRCh38
rs756838913 Chr.15: 66501412-66501412 on Build GRCh38
rs11549578 Chr.15: 66501446-66501446 on Build GRCh38
rs748600371 Chr.15: 66501307-66501309 on Build GRCh38
rs374538654 Chr.15: 66501316-66501316 on Build GRCh38
rs146017997 Chr.15: 66501500-66501500 on Build GRCh38
rs752354297 Chr.15: 66501311-66501311 on Build GRCh38
rs771881549 Chr.15: 66501449-66501449 on Build GRCh38
rs145143124 Chr.15: 66501332-66501332 on Build GRCh38
rs774288053 Chr.15: 66501354-66501354 on Build GRCh38
rs989218263 Chr.15: 66501406-66501406 on Build GRCh38
rs748568380 Chr.15: 66501422-66501422 on Build GRCh38
rs775102412 Chr.15: 66501453-66501453 on Build GRCh38
rs753328340 Chr.15: 66501403-66501403 on Build GRCh38
rs371015044 Chr.15: 66501350-66501350 on Build GRCh38
rs370890435 Chr.15: 66501351-66501351 on Build GRCh38
rs752176433 Chr.15: 66501415-66501415 on Build GRCh38
rs12101954 Chr.15: 66501339-66501339 on Build GRCh38
rs113757438 Chr.15: 66501416-66501416 on Build GRCh38
rs367576894 Chr.15: 66501426-66501426 on Build GRCh38
rs11549596 Chr.15: 66501488-66501488 on Build GRCh38
rs764877683 Chr.15: 66501498-66501498 on Build GRCh38
rs745849000 Chr.15: 66501448-66501448 on Build GRCh38
rs763591992 Chr.15: 66501475-66501475 on Build GRCh38
rs187115320 Chr.15: 66501342-66501342 on Build GRCh38
rs781458060 Chr.15: 66501420-66501420 on Build GRCh38
rs767285238 Chr.15: 66501357-66501357 on Build GRCh38
rs763834786 Chr.15: 66501399-66501399 on Build GRCh38
rs934058990 Chr.15: 66501452-66501452 on Build GRCh38
rs371103900 Chr.15: 66501337-66501337 on Build GRCh38
rs562879783 Chr.15: 66501355-66501355 on Build GRCh38
rs758930136 Chr.15: 66501314-66501314 on Build GRCh38

View Full Product Details