Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38908017-38908248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R16B
Entrez Gene ID
26051
Gene Name
protein phosphatase 1 regulatory subunit 16B
Gene Aliases
ANKRD4, TIMAP
UniGene
Hs.45719
Gene Chromosome Location
Chr.20: 38805693-38923024 on Build GRCh38
Transcripts
NM_001172735.2 NM_015568.3 XM_011528768.2 XM_011528769.2 XM_017027785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4753026 c.1040G>A p.R347Q Chr.20: 38908039-38908039 on Build GRCh38
3963560 c.1191C>A p.D397E Chr.20: 38908190-38908190 on Build GRCh38
4769915 c.1146C>T p.Y382Y Chr.20: 38908145-38908145 on Build GRCh38
1262849 c.1150G>A p.G384R Chr.20: 38908149-38908149 on Build GRCh38
6248509 c.1057G>T p.D353Y Chr.20: 38908056-38908056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764537147 Chr.20: 38908054-38908054 on Build GRCh38
rs780855089 Chr.20: 38908210-38908210 on Build GRCh38
rs539000887 Chr.20: 38908148-38908148 on Build GRCh38
rs374747333 Chr.20: 38908202-38908202 on Build GRCh38
rs142387028 Chr.20: 38908135-38908135 on Build GRCh38
rs754613604 Chr.20: 38908084-38908084 on Build GRCh38
rs754325497 Chr.20: 38908055-38908055 on Build GRCh38
rs201292506 Chr.20: 38908138-38908138 on Build GRCh38
rs868199379 Chr.20: 38908110-38908110 on Build GRCh38
rs753718834 Chr.20: 38908148-38908149 on Build GRCh38
rs371876589 Chr.20: 38908070-38908070 on Build GRCh38
rs944245406 Chr.20: 38908147-38908147 on Build GRCh38
rs137921023 Chr.20: 38908172-38908172 on Build GRCh38
rs117431511 Chr.20: 38908207-38908207 on Build GRCh38
rs768049487 Chr.20: 38908039-38908039 on Build GRCh38
rs140211111 Chr.20: 38908111-38908111 on Build GRCh38
rs563328325 Chr.20: 38908162-38908162 on Build GRCh38
rs1013899924 Chr.20: 38908190-38908190 on Build GRCh38
rs368497752 Chr.20: 38908205-38908205 on Build GRCh38
rs774557229 Chr.20: 38908132-38908132 on Build GRCh38
rs749217242 Chr.20: 38908130-38908130 on Build GRCh38
rs145363233 Chr.20: 38908125-38908125 on Build GRCh38
rs894370197 Chr.20: 38908186-38908186 on Build GRCh38
rs754418915 Chr.20: 38908208-38908208 on Build GRCh38
rs764500209 Chr.20: 38908183-38908183 on Build GRCh38
rs757899810 Chr.20: 38908067-38908067 on Build GRCh38
rs751283329 Chr.20: 38908079-38908079 on Build GRCh38
rs901625179 Chr.20: 38908171-38908171 on Build GRCh38
rs769592966 Chr.20: 38908124-38908124 on Build GRCh38
rs770981411 Chr.20: 38908131-38908131 on Build GRCh38
rs780767201 Chr.20: 38908100-38908100 on Build GRCh38
rs753122696 Chr.20: 38908044-38908044 on Build GRCh38
rs772424757 Chr.20: 38908144-38908144 on Build GRCh38
rs752323046 Chr.20: 38908224-38908224 on Build GRCh38
rs925499759 Chr.20: 38908056-38908056 on Build GRCh38

View Full Product Details