Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47425774-47426035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC3
Entrez Gene ID
5702
Gene Name
proteasome 26S subunit, ATPase 3
Gene Aliases
TBP1
UniGene
Hs.250758
Gene Chromosome Location
Chr.11: 47418769-47426473 on Build GRCh38
Transcripts
NM_002804.4 XM_017018026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033189 c.123C>T p.I41I Chr.11: 47425903-47425903 on Build GRCh38
1981703 c.126C>T p.I42I Chr.11: 47425900-47425900 on Build GRCh38
1981704 c.100C>T p.L34F Chr.11: 47425926-47425926 on Build GRCh38
6757747 c.76-5delT p.? Chr.11: 47425955-47425955 on Build GRCh38
5913264 c.76-6T>A p.? Chr.11: 47425956-47425956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs10838709 Chr.11: 47425795-47425795 on Build GRCh38
rs758236865 Chr.11: 47425954-47425954 on Build GRCh38
rs200093703 Chr.11: 47425841-47425841 on Build GRCh38
rs749766686 Chr.11: 47425817-47425817 on Build GRCh38
rs901888638 Chr.11: 47425959-47425959 on Build GRCh38
rs753723291 Chr.11: 47425873-47425873 on Build GRCh38
rs574933431 Chr.11: 47425903-47425903 on Build GRCh38
rs781614081 Chr.11: 47425960-47425960 on Build GRCh38
rs534528299 Chr.11: 47426008-47426008 on Build GRCh38
rs760200038 Chr.11: 47425858-47425858 on Build GRCh38
rs372362891 Chr.11: 47425853-47425853 on Build GRCh38
rs777845414 Chr.11: 47425992-47425992 on Build GRCh38
rs944532489 Chr.11: 47425860-47425860 on Build GRCh38
rs756297402 Chr.11: 47425987-47425987 on Build GRCh38
rs879192242 Chr.11: 47425820-47425820 on Build GRCh38
rs770530438 Chr.11: 47425961-47425964 on Build GRCh38
rs767194032 Chr.11: 47425851-47425851 on Build GRCh38
rs771206574 Chr.11: 47425828-47425828 on Build GRCh38
rs900279013 Chr.11: 47425947-47425947 on Build GRCh38
rs780543378 Chr.11: 47425964-47425967 on Build GRCh38
rs374507172 Chr.11: 47425861-47425861 on Build GRCh38
rs748336855 Chr.11: 47425979-47425979 on Build GRCh38
rs934612197 Chr.11: 47425812-47425812 on Build GRCh38
rs757158162 Chr.11: 47425894-47425894 on Build GRCh38
rs757697843 Chr.11: 47425845-47425845 on Build GRCh38
rs775097237 Chr.11: 47425857-47425857 on Build GRCh38
rs746712259 Chr.11: 47426012-47426012 on Build GRCh38
rs1052932213 Chr.11: 47425843-47425843 on Build GRCh38
rs750311677 Chr.11: 47425930-47425930 on Build GRCh38
rs749774714 Chr.11: 47425995-47425995 on Build GRCh38
rs771532069 Chr.11: 47426001-47426001 on Build GRCh38

View Full Product Details