Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48587196-48587469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SULT2B1
Entrez Gene ID
6820
Gene Name
sulfotransferase family 2B member 1
Gene Aliases
HSST2
UniGene
Hs.369331
Gene Chromosome Location
Chr.19: 48552172-48599427 on Build GRCh38
Transcripts
NM_004605.2 NM_177973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570572 c.290T>G p.I97S Chr.19: 48587304-48587304 on Build GRCh38
7161549 c.354G>A p.K118K Chr.19: 48587413-48587413 on Build GRCh38
1304863 c.348C>T p.F116F Chr.19: 48587407-48587407 on Build GRCh38
4541144 c.294G>A p.W98* Chr.19: 48587308-48587308 on Build GRCh38
7078916 c.298G>T p.D100Y Chr.19: 48587357-48587357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759460229 Chr.19: 48587422-48587422 on Build GRCh38
rs776667015 Chr.19: 48587442-48587442 on Build GRCh38
rs780820646 Chr.19: 48587360-48587360 on Build GRCh38
rs866043519 Chr.19: 48587296-48587296 on Build GRCh38
rs754602461 Chr.19: 48587356-48587356 on Build GRCh38
rs199730300 Chr.19: 48587223-48587223 on Build GRCh38
rs200535873 Chr.19: 48587275-48587275 on Build GRCh38
rs965219812 Chr.19: 48587443-48587443 on Build GRCh38
rs780615173 Chr.19: 48587343-48587343 on Build GRCh38
rs757436486 Chr.19: 48587299-48587299 on Build GRCh38
rs774401100 Chr.19: 48587418-48587418 on Build GRCh38
rs765525281 Chr.19: 48587445-48587445 on Build GRCh38
rs768213131 Chr.19: 48587245-48587245 on Build GRCh38
rs893577303 Chr.19: 48587226-48587226 on Build GRCh38
rs932416497 Chr.19: 48587318-48587318 on Build GRCh38
rs1004344891 Chr.19: 48587359-48587359 on Build GRCh38
rs749117260 Chr.19: 48587378-48587378 on Build GRCh38
rs773038899 Chr.19: 48587264-48587264 on Build GRCh38
rs149116557 Chr.19: 48587291-48587291 on Build GRCh38
rs747459444 Chr.19: 48587348-48587348 on Build GRCh38
rs200817158 Chr.19: 48587270-48587270 on Build GRCh38
rs373332265 Chr.19: 48587444-48587444 on Build GRCh38
rs769753162 Chr.19: 48587252-48587252 on Build GRCh38
rs986181196 Chr.19: 48587401-48587401 on Build GRCh38
rs770576995 Chr.19: 48587383-48587383 on Build GRCh38
rs558429244 Chr.19: 48587364-48587364 on Build GRCh38
rs751390677 Chr.19: 48587286-48587286 on Build GRCh38
rs1027668633 Chr.19: 48587372-48587372 on Build GRCh38
rs764196414 Chr.19: 48587433-48587433 on Build GRCh38
rs1052124 Chr.19: 48587344-48587344 on Build GRCh38
rs140526640 Chr.19: 48587246-48587246 on Build GRCh38
rs372325021 Chr.19: 48587292-48587292 on Build GRCh38
rs777927515 Chr.19: 48587355-48587355 on Build GRCh38
rs769594886 Chr.19: 48587370-48587370 on Build GRCh38
rs765637140 Chr.19: 48587300-48587300 on Build GRCh38
rs527454384 Chr.19: 48587232-48587232 on Build GRCh38
rs201511288 Chr.19: 48587342-48587342 on Build GRCh38
rs367706728 Chr.19: 48587233-48587233 on Build GRCh38
rs757672172 Chr.19: 48587222-48587222 on Build GRCh38
rs138292714 Chr.19: 48587313-48587313 on Build GRCh38
rs746398875 Chr.19: 48587229-48587229 on Build GRCh38
rs377120502 Chr.19: 48587373-48587373 on Build GRCh38
rs200176089 Chr.19: 48587297-48587297 on Build GRCh38

View Full Product Details