Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11760774-11760999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL3
Entrez Gene ID
10943
Gene Name
male-specific lethal 3 homolog (Drosophila)
Gene Aliases
MSL3L1
UniGene
Hs.655288
Gene Chromosome Location
Chr.X: 11758159-11775753 on Build GRCh38
Transcripts
NM_001193270.2 NM_001282174.1 NM_006800.3 NM_078628.1 NM_078629.3 XM_005274440.2 XM_011545442.2

SNP Detail

SNP ID Chromosome Location
rs746226340 Chr.X: 11760824-11760824 on Build GRCh38
rs368156652 Chr.X: 11760965-11760965 on Build GRCh38
rs764860597 Chr.X: 11760902-11760902 on Build GRCh38
rs772930018 Chr.X: 11760903-11760903 on Build GRCh38
rs779486885 Chr.X: 11760818-11760818 on Build GRCh38
rs147813916 Chr.X: 11760847-11760847 on Build GRCh38
rs778282766 Chr.X: 11760804-11760804 on Build GRCh38
rs757486008 Chr.X: 11760809-11760809 on Build GRCh38
rs191141351 Chr.X: 11760862-11760862 on Build GRCh38
rs897477398 Chr.X: 11760864-11760864 on Build GRCh38
rs757550557 Chr.X: 11760941-11760941 on Build GRCh38
rs766049541 Chr.X: 11760921-11760921 on Build GRCh38
rs762777627 Chr.X: 11760913-11760913 on Build GRCh38
rs377114479 Chr.X: 11760859-11760859 on Build GRCh38
rs749597577 Chr.X: 11760866-11760866 on Build GRCh38
rs754903127 Chr.X: 11760972-11760972 on Build GRCh38
rs372603177 Chr.X: 11760894-11760894 on Build GRCh38
rs761524573 Chr.X: 11760899-11760899 on Build GRCh38
rs867279259 Chr.X: 11760844-11760844 on Build GRCh38
rs768171462 Chr.X: 11760863-11760863 on Build GRCh38
rs745839076 Chr.X: 11760853-11760855 on Build GRCh38

View Full Product Details