Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54982514-54982765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP2
Entrez Gene ID
55655
Gene Name
NLR family pyrin domain containing 2
Gene Aliases
CLR19.9, NALP2, NBS1, PAN1, PYPAF2
UniGene
Hs.369279
Gene Chromosome Location
Chr.19: 54965284-55001142 on Build GRCh38
Transcripts
NM_001174081.1 NM_001174082.1 NM_001174083.1 NM_017852.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6727131 c.867C>A p.G289G Chr.19: 54982565-54982565 on Build GRCh38
4512459 c.903C>T p.I301I Chr.19: 54982601-54982601 on Build GRCh38
5856764 c.983C>T p.P328L Chr.19: 54982681-54982681 on Build GRCh38
4991343 c.1008G>C p.T336T Chr.19: 54982706-54982706 on Build GRCh38
6152279 c.939G>C p.P313P Chr.19: 54982637-54982637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778010743 Chr.19: 54982597-54982597 on Build GRCh38
rs747472514 Chr.19: 54982599-54982599 on Build GRCh38
rs754777768 Chr.19: 54982588-54982588 on Build GRCh38
rs371910953 Chr.19: 54982650-54982650 on Build GRCh38
rs770490629 Chr.19: 54982682-54982682 on Build GRCh38
rs561774583 Chr.19: 54982644-54982644 on Build GRCh38
rs761378450 Chr.19: 54982603-54982603 on Build GRCh38
rs768813006 Chr.19: 54982624-54982626 on Build GRCh38
rs756436723 Chr.19: 54982667-54982667 on Build GRCh38
rs760216362 Chr.19: 54982624-54982624 on Build GRCh38
rs200257622 Chr.19: 54982540-54982540 on Build GRCh38
rs144397371 Chr.19: 54982601-54982601 on Build GRCh38
rs149735961 Chr.19: 54982708-54982708 on Build GRCh38
rs766120038 Chr.19: 54982584-54982584 on Build GRCh38
rs769589997 Chr.19: 54982737-54982737 on Build GRCh38
rs771069040 Chr.19: 54982546-54982547 on Build GRCh38
rs755393462 Chr.19: 54982707-54982707 on Build GRCh38
rs765030338 Chr.19: 54982686-54982686 on Build GRCh38
rs377520886 Chr.19: 54982690-54982690 on Build GRCh38
rs772739635 Chr.19: 54982618-54982618 on Build GRCh38
rs777161045 Chr.19: 54982562-54982562 on Build GRCh38
rs756602998 Chr.19: 54982704-54982704 on Build GRCh38
rs188819279 Chr.19: 54982614-54982614 on Build GRCh38
rs865812246 Chr.19: 54982623-54982623 on Build GRCh38
rs146500465 Chr.19: 54982652-54982652 on Build GRCh38
rs76399676 Chr.19: 54982539-54982539 on Build GRCh38
rs148817929 Chr.19: 54982743-54982743 on Build GRCh38
rs767083504 Chr.19: 54982577-54982577 on Build GRCh38
rs564394457 Chr.19: 54982587-54982587 on Build GRCh38
rs200189881 Chr.19: 54982689-54982689 on Build GRCh38
rs771360716 Chr.19: 54982554-54982554 on Build GRCh38
rs577834121 Chr.19: 54982619-54982619 on Build GRCh38
rs141427711 Chr.19: 54982636-54982636 on Build GRCh38
rs139921503 Chr.19: 54982662-54982662 on Build GRCh38
rs1045014130 Chr.19: 54982732-54982732 on Build GRCh38
rs3745904 Chr.19: 54982602-54982602 on Build GRCh38
rs199475713 Chr.19: 54982718-54982718 on Build GRCh38
rs866811421 Chr.19: 54982664-54982664 on Build GRCh38
rs776561473 Chr.19: 54982684-54982684 on Build GRCh38
rs145646456 Chr.19: 54982710-54982710 on Build GRCh38
rs772899546 Chr.19: 54982672-54982672 on Build GRCh38
rs781762168 Chr.19: 54982649-54982649 on Build GRCh38
rs761373612 Chr.19: 54982576-54982576 on Build GRCh38
rs767499946 Chr.19: 54982692-54982692 on Build GRCh38
rs775549835 Chr.19: 54982740-54982740 on Build GRCh38
rs755257910 Chr.19: 54982541-54982541 on Build GRCh38
rs748235569 Chr.19: 54982548-54982548 on Build GRCh38
rs776824462 Chr.19: 54982610-54982612 on Build GRCh38
rs61735080 Chr.19: 54982637-54982637 on Build GRCh38
rs138537360 Chr.19: 54982563-54982563 on Build GRCh38
rs780650039 Chr.19: 54982705-54982705 on Build GRCh38
rs779043525 Chr.19: 54982543-54982543 on Build GRCh38
rs147098165 Chr.19: 54982553-54982553 on Build GRCh38
rs753776748 Chr.19: 54982635-54982635 on Build GRCh38
rs139643111 Chr.19: 54982643-54982643 on Build GRCh38
rs1019187055 Chr.19: 54982674-54982674 on Build GRCh38
rs193065675 Chr.19: 54982703-54982703 on Build GRCh38
rs149627959 Chr.19: 54982594-54982594 on Build GRCh38
rs375621121 Chr.19: 54982688-54982688 on Build GRCh38
rs144092754 Chr.19: 54982647-54982647 on Build GRCh38
rs746508494 Chr.19: 54982680-54982680 on Build GRCh38
rs779179826 Chr.19: 54982593-54982593 on Build GRCh38
rs773925733 Chr.19: 54982574-54982574 on Build GRCh38
rs749628445 Chr.19: 54982706-54982706 on Build GRCh38
rs766281794 Chr.19: 54982631-54982631 on Build GRCh38
rs369991898 Chr.19: 54982583-54982583 on Build GRCh38
rs745776240 Chr.19: 54982731-54982731 on Build GRCh38
rs370946962 Chr.19: 54982613-54982613 on Build GRCh38
rs747795316 Chr.19: 54982671-54982671 on Build GRCh38
rs759402397 Chr.19: 54982685-54982685 on Build GRCh38
rs752904721 Chr.19: 54982641-54982641 on Build GRCh38
rs373171651 Chr.19: 54982595-54982595 on Build GRCh38

View Full Product Details