Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45825602-45825855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPG5
Entrez Gene ID
57724
Gene Name
ectopic P-granules autophagy protein 5 homolog
Gene Aliases
HEEW1, KIAA1632, VICIS
UniGene
Hs.514843
Gene Chromosome Location
Chr.18: 45800590-45967339 on Build GRCh38
Transcripts
XM_017025890.1 NM_213602.2

Gene Symbol
SIGLEC15
Entrez Gene ID
284266
Gene Name
sialic acid binding Ig like lectin 15
Gene Aliases
CD33L3, HsT1361, SIGLEC-15
UniGene
Hs.287692
Gene Chromosome Location
Chr.18: 45825580-45842556 on Build GRCh38
Transcripts
XM_017025890.1 NM_213602.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342255 c.47C>T p.P16L Chr.18: 45825775-45825775 on Build GRCh38
7429743 c.2T>C p.M1T Chr.18: 45825730-45825730 on Build GRCh38
2877507 c.19C>T p.L7L Chr.18: 45825747-45825747 on Build GRCh38
267424 c.40G>A p.V14I Chr.18: 45825768-45825768 on Build GRCh38
5940809 c.17G>C p.W6S Chr.18: 45825745-45825745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768210022 Chr.18: 45825776-45825776 on Build GRCh38
rs757655809 Chr.18: 45825814-45825814 on Build GRCh38
rs142641185 Chr.18: 45825747-45825747 on Build GRCh38
rs765480921 Chr.18: 45825802-45825802 on Build GRCh38
rs929511773 Chr.18: 45825639-45825639 on Build GRCh38
rs748907923 Chr.18: 45825775-45825775 on Build GRCh38
rs762630612 Chr.18: 45825689-45825689 on Build GRCh38
rs780441124 Chr.18: 45825683-45825683 on Build GRCh38
rs375991022 Chr.18: 45825684-45825684 on Build GRCh38
rs750983843 Chr.18: 45825819-45825819 on Build GRCh38
rs751882312 Chr.18: 45825751-45825751 on Build GRCh38
rs761546128 Chr.18: 45825786-45825786 on Build GRCh38
rs774025905 Chr.18: 45825695-45825695 on Build GRCh38
rs199664451 Chr.18: 45825767-45825767 on Build GRCh38
rs115113388 Chr.18: 45825790-45825790 on Build GRCh38
rs775160452 Chr.18: 45825686-45825686 on Build GRCh38
rs139939061 Chr.18: 45825760-45825760 on Build GRCh38
rs752724402 Chr.18: 45825811-45825811 on Build GRCh38
rs970458308 Chr.18: 45825820-45825820 on Build GRCh38
rs150984553 Chr.18: 45825764-45825764 on Build GRCh38
rs758684969 Chr.18: 45825748-45825748 on Build GRCh38
rs536100309 Chr.18: 45825813-45825813 on Build GRCh38
rs763176236 Chr.18: 45825812-45825812 on Build GRCh38
rs779486475 Chr.18: 45825830-45825830 on Build GRCh38
rs372096305 Chr.18: 45825742-45825742 on Build GRCh38
rs549055099 Chr.18: 45825636-45825636 on Build GRCh38
rs201025712 Chr.18: 45825797-45825797 on Build GRCh38
rs1012211399 Chr.18: 45825650-45825650 on Build GRCh38
rs745403677 Chr.18: 45825683-45825683 on Build GRCh38
rs200364769 Chr.18: 45825696-45825696 on Build GRCh38
rs192098402 Chr.18: 45825818-45825818 on Build GRCh38
rs1022514725 Chr.18: 45825704-45825704 on Build GRCh38
rs369769268 Chr.18: 45825690-45825690 on Build GRCh38
rs371157596 Chr.18: 45825658-45825658 on Build GRCh38
rs999847666 Chr.18: 45825756-45825756 on Build GRCh38
rs778132336 Chr.18: 45825750-45825750 on Build GRCh38
rs767588812 Chr.18: 45825707-45825707 on Build GRCh38
rs1440822 Chr.18: 45825827-45825827 on Build GRCh38
rs753931063 Chr.18: 45825709-45825709 on Build GRCh38
rs567556732 Chr.18: 45825640-45825640 on Build GRCh38
rs867045623 Chr.18: 45825778-45825778 on Build GRCh38
rs375713508 Chr.18: 45825799-45825799 on Build GRCh38
rs372386663 Chr.18: 45825798-45825798 on Build GRCh38
rs199680905 Chr.18: 45825723-45825723 on Build GRCh38
rs529088073 Chr.18: 45825763-45825763 on Build GRCh38
rs376022672 Chr.18: 45825665-45825665 on Build GRCh38

View Full Product Details