Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35432530-35432757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
UniGene
Hs.55044
Gene Chromosome Location
Chr.11: 35431827-35530300 on Build GRCh38
Transcripts
NM_001001991.2 NM_001282675.1 NM_001282676.1 NM_015430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4819218 c.1984G>A p.D662N Chr.11: 35432586-35432586 on Build GRCh38
1353680 c.1893C>T p.N631N Chr.11: 35432677-35432677 on Build GRCh38
1235749 c.1894G>A p.D632N Chr.11: 35432676-35432676 on Build GRCh38
6242647 c.2022T>C p.P674P Chr.11: 35432548-35432548 on Build GRCh38
354984 c.2021C>A p.P674H Chr.11: 35432549-35432549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376867555 Chr.11: 35432576-35432576 on Build GRCh38
rs769215931 Chr.11: 35432681-35432681 on Build GRCh38
rs146058819 Chr.11: 35432599-35432599 on Build GRCh38
rs894822042 Chr.11: 35432693-35432693 on Build GRCh38
rs767974456 Chr.11: 35432602-35432602 on Build GRCh38
rs777304316 Chr.11: 35432692-35432692 on Build GRCh38
rs146387977 Chr.11: 35432575-35432575 on Build GRCh38
rs779803306 Chr.11: 35432553-35432553 on Build GRCh38
rs758102474 Chr.11: 35432634-35432634 on Build GRCh38
rs368802780 Chr.11: 35432667-35432667 on Build GRCh38
rs776078670 Chr.11: 35432639-35432639 on Build GRCh38
rs138681708 Chr.11: 35432617-35432617 on Build GRCh38
rs147203964 Chr.11: 35432737-35432737 on Build GRCh38
rs747015590 Chr.11: 35432557-35432557 on Build GRCh38
rs755000531 Chr.11: 35432646-35432646 on Build GRCh38
rs756828776 Chr.11: 35432616-35432616 on Build GRCh38
rs775832642 Chr.11: 35432607-35432607 on Build GRCh38
rs61744525 Chr.11: 35432560-35432560 on Build GRCh38
rs766074036 Chr.11: 35432711-35432711 on Build GRCh38
rs749326535 Chr.11: 35432655-35432655 on Build GRCh38
rs201670870 Chr.11: 35432676-35432676 on Build GRCh38
rs115205961 Chr.11: 35432649-35432649 on Build GRCh38
rs61738938 Chr.11: 35432650-35432650 on Build GRCh38
rs761316229 Chr.11: 35432608-35432608 on Build GRCh38
rs993501857 Chr.11: 35432600-35432600 on Build GRCh38
rs979867291 Chr.11: 35432592-35432592 on Build GRCh38
rs149354945 Chr.11: 35432638-35432638 on Build GRCh38
rs554767403 Chr.11: 35432738-35432738 on Build GRCh38
rs774588409 Chr.11: 35432594-35432594 on Build GRCh38
rs1004210602 Chr.11: 35432680-35432680 on Build GRCh38
rs201062106 Chr.11: 35432677-35432677 on Build GRCh38
rs758362043 Chr.11: 35432628-35432629 on Build GRCh38
rs148541461 Chr.11: 35432700-35432700 on Build GRCh38
rs749497491 Chr.11: 35432578-35432578 on Build GRCh38
rs199759649 Chr.11: 35432666-35432666 on Build GRCh38
rs753344815 Chr.11: 35432613-35432613 on Build GRCh38
rs750109884 Chr.11: 35432626-35432626 on Build GRCh38
rs763687291 Chr.11: 35432609-35432609 on Build GRCh38
rs193167829 Chr.11: 35432727-35432727 on Build GRCh38
rs769729805 Chr.11: 35432567-35432567 on Build GRCh38
rs145222740 Chr.11: 35432701-35432701 on Build GRCh38
rs762732343 Chr.11: 35432705-35432705 on Build GRCh38
rs771075275 Chr.11: 35432585-35432585 on Build GRCh38
rs754799699 Chr.11: 35432731-35432731 on Build GRCh38
rs781059398 Chr.11: 35432733-35432733 on Build GRCh38
rs567309396 Chr.11: 35432605-35432605 on Build GRCh38

View Full Product Details